Merck
HomeProductsMaterials ScienceEnergy MaterialsSolution & Vapor Deposition Precursors
Solution & Vapor Deposition Precursors
Graphic of solution deposition and vapor deposition precusors
Chemical solution deposition and chemical vapor deposition are two powerful techniques used to form high-quality and precision thin solid films and coatings. Solution deposition, also known as sol-gel processing, is a popular method used to prepare a wide range of inorganic and hybrid composite materials from precursor solutions. A colloidal suspension known as “sol” is generated, converted to a gel, and subsequently transitions into a solid. However, vapor deposition uses a host of techniques to convert and employ precursors or target materials in the gaseous phase to form engineered films on substrates. Our selection of products for deposition techniques allows you to precisely tailor thin films and their coating properties. Explore our broad range of high-quality and dependable thin film deposition precursor chemicals as per your chosen method and application.
Read more about
Products
Sort by Relevance
Showing 1-20 of 330
Change View
Showing 1-20 of 330
Compare
Product Number
Product Name
Product Description
Pricing
Solution Deposition Precursors
We offer a diverse array of dedicated solution deposition precursors. They are available with 55 different base metals as well as acetate, acetylacetonate, tert-butoxide, isopropoxide, phenoxide, ethoxide, tri-sec-butoxide, methoxide, and 2-ethylhexanoate along with a variety of other functionalities. These products are offered in purities ranging from 90% to 99.999%, in assorted concentrations in select solvents, and with specific hydrated forms to assist your distinct chemistry.
Chemical Vapor Deposition (CVD) Precursors/Atomic Layer Deposition (ALD) Precursors
We provide high-quality volatile organometallic, metal, and metalorganic precursors for CVD/ALD. For convenience and safety, the prepackaged precursors come in steel cylinders for use with an assortment of deposition systems.
Physical Vapor Deposition Materials
Physical vapor deposition (PVD) utilizes vaporized material from solid source material to deposit thin films on a substrate. We offer high-purity sputtering targets, pellets, metal foils, and evaporation slugs for use in various PVD applications. This includes microelectronic devices, battery electrodes, diffusion barriers, and optical coatings.
Related Product Resources
  • Deposition Grade Silanes for Sol-Gel Processes
    Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.
  • Materials for Thin Film Solid Oxide Fuel Cells (TF-SOFCs)
    This technical spotlight focuses on the thin film solid oxide fuel cells (TF-SOFC). TF-SOFCs refer to solid oxide fuel cells with electrolyte thin films. This review highlights technical advantages of TF-SOFC and introduces deposition methods of electrolyte thin films to fabricate TF-SOFC.
  • Boron Nitride Nanotubes: Properties, Synthesis and Applications
    Boron nitride nanotubes (BNNT) are close structural analogs of carbon nanotubes (CNT), which are high aspect ratio nanotubular material, where carbon atoms are alternately substituted by nitrogen and boron atoms.
  • CoMoCAT® Single-wall Carbon Nanotubes
    The CoMoCAT® method of single-walled carbon nanotube (SWNT) synthesis yields high purity SWNTs with specific chiralities and narrow distributions of tube diameters.