All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH(CH3)CH(CH3)2
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696869
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.404 (lit.)
bp
113-114 °C (lit.)
mp
−110 °C (lit.)
density
0.719 g/mL at 25 °C (lit.)
SMILES string
CC(C)C(C)C(C)C
InChI
1S/C8H18/c1-6(2)8(5)7(3)4/h6-8H,1-5H3
InChI key
RLPGDEORIPLBNF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Kinetics and mechanism of reaction of OH radicals with 2,3,4-trimethylpentane in the presence of NO has been investigated. Kinetics of reactions of C6H5 radical with 2,3,4-trimethylpentane has been measured by cavity ringdown spectrometry.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.2 °F - closed cup
Flash Point(C)
4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lack of detectable metabolism for solubilized 2,3,4-trimethylpentane by rat kidney proximal tubules.
N J DelRaso et al.
Toxicology letters, 54(2-3), 337-344 (1990-12-01)
Primary proximal tubule suspension cultures exposed to solubilized 2,3,4-trimethylpentane (2,3,4-TMP) resulted in a linear dose response, as determined by cellular lactate dehydrogenase leakage. The EC50 for 2,3,4-TMP was 16.3 mM. Metabolite analysis by gas chromatography/mass spectrometry of supernate and cell
Sara M Aschmann et al.
Environmental science & technology, 38(19), 5038-5045 (2004-10-28)
Alkanes are important constituents of gasoline fuel and vehicle exhaust, with branched alkanes comprising a significant fraction of the total alkanes observed in urban areas. Using a relative rate method, a rate constant for the reaction of OH radicals with
K O Yu et al.
Biomedical & environmental mass spectrometry, 14(11), 639-641 (1987-11-01)
Branched-chain hydrocarbon-induced nephropathy in male rats was examined using 2,3,4-trimethylpentane. Lesions are elected only in male rats, not in females or in controls. Mechanisms of nephropathy may be the interaction of metabolites with the male rat-specific protein alpha 2u globulin.
Kinetics of C~ 6H~ 5 Radical Reactions with 2-Methylpropane, 2, 3-Dimethylbutane and 2, 3, 4-Trimethylpentane.
Park J, et al.
International Journal of Chemical Kinetics, 31(9), 645-653 (1999)
N J Delraso et al.
In vitro cellular & developmental biology : journal of the Tissue Culture Association, 25(11), 1031-1038 (1989-11-01)
Primary rat hepatocyte suspension cultures (approximately 2 X 10(6) cells) exposed to solubilized 2,3,4-trimethylpentane at concentrations ranging from 7.9 to 31.5 mM under two different culture conditions resulted in a linear dose response, as determined by lactate dehydrogenase leakage and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service