Skip to Content
Merck
All Photos(1)

Key Documents

PHR1915

Supelco

2,2,4-Trimethylpentane (Isooctane)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,2,4-Trimethylpentane, Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

CofA

current certificate can be downloaded

autoignition temp.

745 °F

expl. lim.

6 %

packaging

ampule of 3x1.2 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 2,2,4-Trimethylpentane (Isooctane) is a primary reference fuel with an arbitrary octane number of 100 as it burns smoothly without resulting in any knocking. It is mainly used to improve the octane rating of combustion fuels.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB0908 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nurettin Sahiner et al.
Carbohydrate polymers, 159, 29-38 (2017-01-01)
Poly(hyaluronic acid) (p(HA)) particles with sizes from few hundred nm to few tens of micrometer were synthesized by using epoxy groups containing crosslinker glycerol diglycidyl ether (GDE) with high yield, 94±5%. P(HA) particles were oxidized by treatment with sodium periodate
Amirhossein Hakamivala et al.
Bioactive materials, 5(1), 142-152 (2020-02-20)
Cartilage injury affects millions of people throughout the world, and at this time there is no cure. While transplantation of stem cells has shown some success in the treatment of injured cartilage, such treatment is limited by limited cell sources

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service