Skip to Content
Merck
All Photos(3)

Documents

283096

Sigma-Aldrich

tert-Amyl methyl ether

97%

Synonym(s):

1,1-Dimethylpropyl methyl ether, Methyl 2-methyl-2-butyl ether, Methyl tert-pentyl ether, TAME

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2OCH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.389 (lit.)
n20/D 1.3896

bp

85-86 °C (lit.)

density

0.76-0.78 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)OC

InChI

1S/C6H14O/c1-5-6(2,3)7-4/h5H2,1-4H3

InChI key

HVZJRWJGKQPSFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

tert-Amyl methyl ether is an fuel ether oxygenate and has been quantitated in water by membrane extraction ion mobility spectrometry. tert-Amyl methyl ether is also referred to as 2-methoxy-2-methyl butane. Kinetics of ion exchange resin (Amberlyst 18) catalyzed tert-amyl methyl ether reactions have been investigated.

Application


  • Identification of environmental hazards of gasoline oxygenate tert-amyl methyl ether (TAME).: This study evaluates the environmental risks associated with the use of tert-Amyl methyl ether (TAME) as a gasoline oxygenate. The research highlights potential hazards to environmental and human health, underscoring the need for careful assessment and regulation of TAME use in fuels. (Huttunen et al., 1997).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanna Holopainen et al.
Talanta, 106, 448-453 (2013-04-20)
Fuel oxygenates are environmentally detrimental compounds due to their rapid migration to groundwater. Fuel oxygenates have been reported to cause taste and odour problems in drinking water, and they also have long-term health effects. Feasible analytical methods are required to
Kinetics of heterogeneously catalyzed tert-amyl methyl ether reactions in the liquid phase.
Rihko LK and Krause AOI.
Industrial & Engineering Chemistry Research, 34(4), 1172-1180 (1995)
James E Landmeyer et al.
Ground water, 48(1), 30-41 (2009-08-12)
Groundwater contamination by fuel-related compounds such as the fuel oxygenates methyl tert-butyl ether (MTBE), tert-butyl alcohol (TBA), and tert-amyl methyl ether (TAME) presents a significant issue to managers and consumers of groundwater and surface water that receives groundwater discharge. Four
R W Tyl et al.
Journal of applied toxicology : JAT, 23(6), 397-410 (2003-11-25)
Under Office of Prevention, Pesticides and Toxic Substances draft guidelines, CD weanling F0 rats (30 of each gender per group) inhaled tertiary amyl methyl ether vapor at 0, 250, 1500 or 3000 ppm 5 days a week and 6 h
Dorothea M Kujawinski et al.
Journal of environmental monitoring : JEM, 12(1), 347-354 (2010-01-19)
In order to monitor the behaviour of contaminants in the aqueous environment effective enrichment techniques often have to be employed due to their low concentrations. In this work a robust and sensitive purge and trap-gas chromatography-isotope ratio mass spectrometry method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service