Skip to Content
Merck
All Photos(3)

Key Documents

254614

Sigma-Aldrich

N-(Phenylseleno)phthalimide

technical grade

Synonym(s):

2-(Phenylseleno)-1H-isoindole-1,3(2H)-dione, 2-Phenylselanylisoindole-1,3-dione, N-Phenylselenylphthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2Se
CAS Number:
Molecular Weight:
302.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

mp

181-184 °C (lit.)

storage temp.

−20°C

SMILES string

O=C1N([Se]c2ccccc2)C(=O)c3ccccc13

InChI

1S/C14H9NO2Se/c16-13-11-8-4-5-9-12(11)14(17)15(13)18-10-6-2-1-3-7-10/h1-9H

InChI key

TYZFQSLJTWPSDS-UHFFFAOYSA-N

Application

N-(Phenylseleno)phthalimide is a versatile selenamide reagent and has been used:
  • to selectively derivatize thiols for mass spectrometric analysis
  • to derivatize thiol peptides in protein digests
  • in L-prolinamide or pyrrolidine trifluoromethanesulfonamide promoted α-selenenylation reactions of aldehydes and ketones
  • for introducing PhSe into protected glycals, ketones, aldehydes, stannanes and exocyclic alkenes

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 7755-7755 (1993)
Tetrahedron Letters, 33, 6243-6243 (1992)
Synlett, 419-419 (1992)
Tetrahedron Letters, 34, 7915-7915 (1993)
Jian Wang et al.
The Journal of organic chemistry, 70(14), 5678-5687 (2005-07-02)
[reaction: see text] A new catalytic method for direct alpha-selenenylation reactions of aldehydes and ketones has been developed. The results of exploratory studies have demonstrated that L-prolinamide is an effective catalyst for alpha-selenenylation reactions of aldehydes, whereas pyrrolidine trifluoromethanesulfonamide efficiently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service