Skip to Content
Merck
All Photos(4)

Key Documents

S1650

Sigma-Aldrich

Seleno-DL-cystine

>98% (TLC), suitable for HPLC

Synonym(s):

3,3′-diselenobis- alanine,

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O4Se2
CAS Number:
Molecular Weight:
334.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

Seleno-DL-cystine, powder

Assay

>98% (TLC)

Quality Level

form

powder

technique(s)

HPLC: suitable

mp

222 °C

storage temp.

−20°C

SMILES string

NC(C[Se][Se]CC(N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4Se2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)

InChI key

JULROCUWKLNBSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Burguière et al.
Journal of bacteriology, 186(15), 4875-4884 (2004-07-21)
The symporter YhcL and two ATP binding cassette transporters, YtmJKLMN and YckKJI, were shown to mediate L-cystine uptake in Bacillus subtilis. A triple DeltayhcL DeltaytmJKLMN DeltayckK mutant was unable to grow in the presence of L-cystine and to take up
Mamoru Haratake et al.
Bioconjugate chemistry, 19(9), 1831-1839 (2008-08-19)
We synthesized nanoparticulate glutathione peroxidase (GPx) mimics in which selenocystine (SeCyst) was conjugated to a hydrophilic linear polysaccharide, pullulan (Pul). The SeCyst ester-conjugated Pul derivatives (SeCyst-Pul) in phosphate buffer (pH 7) were treated with a sonicator to spontaneously form particulate
Thomas Hofer et al.
Biochemistry, 48(50), 12047-12057 (2009-11-10)
Antibody conjugates have broad utility in basic, preclinical, and clinical applications. Conventional antibody conjugation through the amine group of lysine or the thiol group of cysteine residues yields heterogeneous products of undefined stoichiometry and considerable batch-to-batch variability. To preserve the
Tianfeng Chen et al.
Journal of agricultural and food chemistry, 56(22), 10574-10581 (2008-10-31)
Selenocystine (SeC) is a nutritionally available selenoamino acid with selective anticancer effects on a number of human cancer cell lines. The present study shows that SeC inhibited the proliferation of human breast adenocarcinoma MCF-7 cells in a time- and dose-dependent
Spela Mechora et al.
Chemosphere, 84(11), 1636-1641 (2011-06-28)
The uptake of Se (VI) by two aquatic plants, Myriophyllum spicatum L. and Ceratophyllum demersum L., and its effects on their physiological characteristics have been studied. Plants were cultivated outdoors under semi-controlled conditions and in two concentrations of Na selenate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service