Skip to Content
Merck
All Photos(1)

Documents

251860

Sigma-Aldrich

1-Iodononane

95%

Synonym(s):

Nonyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8I
CAS Number:
Molecular Weight:
254.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.487 (lit.)

bp

107-108 °C/8 mmHg (lit.)

density

1.288 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCI

InChI

1S/C9H19I/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3

InChI key

OGSJMFCWOUHXHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Iodononane has been used in the synthesis of:
  • Sch II, a cerebroside that induces fruiting body formation in the basidiomycete Schizophyllum commune
  • 2,6-dimethyl-4-nonylpyridine
  • 2-tridecanone

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound mediated synthesis of a few naturally occurring compounds.
Singh J, et al.
Indian J. Chem. B, 40(5), 386-390 (2005)
Fiona J Black et al.
Organic & biomolecular chemistry, 8(5), 1188-1193 (2010-02-19)
(4E,8E,10E)-9-Methyl-4,8,10-sphingatrienine, a core component of marine sphingolipids, was synthesised for the first time using a copper(I)-mediated 1,2-metallate rearrangement of a lithiated glycal as a key step. It was converted to phalluside-1, a cerebroside isolated from the ascidian Phallusia fumigate. By
Sulfone-mediated synthesis of polysubstituted pyridines.
Craig D and Henry GD.
Tetrahedron Letters, 46(15), 2559-2562 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service