Skip to Content
Merck
All Photos(1)

Key Documents

275352

Sigma-Aldrich

2-Hydroxy-5-nitrobenzaldehyde

98%

Synonym(s):

5-Nitrosalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(NO2)CHO
CAS Number:
Molecular Weight:
167.12
Beilstein:
512565
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

125-128 °C (lit.)

functional group

aldehyde
nitro

SMILES string

[H]C(=O)c1cc(ccc1O)[N+]([O-])=O

InChI

1S/C7H5NO4/c9-4-5-3-6(8(11)12)1-2-7(5)10/h1-4,10H

InChI key

IHFRMUGEILMHNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxy-5-nitrobenzaldehyde is a nitroaromatic compound used to prepare Schiff base ligands.

The interaction of 2-hydroxy-5-nitrobenzaldehyde and chlorogenic acid (CHL) with the components of the rat hepatic glucose 6-phosphatase system was studied.

Physical properties

Free of 3-nitro isomer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Van Noorden et al.
The Histochemical journal, 19(9), 483-487 (1987-09-01)
The histochemical fluorescence method using 5-nitrosalicylaldehyde for the demonstration of cathepsin B activity has been used. Precipitation of the fluorescent final reaction product was analysed continuously during incubation for cathepsin B activity. Unfixed cultured human fibroblasts as well as cryostat
Anda-Mihaela Olaru et al.
Carbohydrate polymers, 179, 59-70 (2017-11-08)
A series of hydrogels based on chitosan polyamine and nitrosalicylaldehyde were prepared via dynamic covalent chemistry (DCC), by imination and transimination reactions towards ordered clusters which play the role of crosslinking nodes of the chitosan network. The hydrogelation mechanism has
Anas G Elsafy et al.
Sensors (Basel, Switzerland), 18(7) (2018-07-13)
(E)-2-((benzo[d]thiazol-2-ylimino)methyl)-4-nitrophenol 1 and (E)-2-(((6-methoxybenzo[d]thiazol-2-yl)imino)methyl)-4-nitrophenol 2 were synthesized efficiently under microwave conditions. The structures were confirmed using IR, ¹H NMR, and 13C NMR. UV-vis. Fluorescence investigations demonstrated that 1 and 2 are sensitive and selective sensors for detection of cyanide over
W J Arion et al.
Archives of biochemistry and biophysics, 351(2), 279-285 (1998-04-29)
S 3483, a synthetic derivative of chlorogenic acid (CHL), was found to be a reversible, linear competitive inhibitor of the glucose-6-phosphatase (Glc-6-Pase) system in rat renal microsomes and rat and human liver microsomes. The Ki for S 3483 in rat
Melwin Colaço et al.
ChemistryOpen, 7(7), 520-526 (2018-07-14)
Direct access to the protonated merocyanine forms of two substituted spiropyrans by mechanosynthesis or sonochemistry was explored. The compounds were formed by the condensation reaction of the methyleneindolium iodide salt with salicylaldehyde derivatives. X-ray crystallography, 1H NMR spectroscopy, ab initio geometry optimization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service