Skip to Content
Merck
All Photos(2)

Key Documents

202851

Sigma-Aldrich

Boric anhydride

99.999% trace metals basis

Synonym(s):

Boron trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2O3
CAS Number:
Molecular Weight:
69.62
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38120103
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

Quality Level

Assay

99.999% trace metals basis

form

crystals or chunks

reaction suitability

reagent type: catalyst
core: boron

mp

450 °C (lit.)

solubility

ethanol: soluble at 20 °C

density

2.46 g/mL at 25 °C (lit.)

SMILES string

O=BOB=O

InChI

1S/B2O3/c3-1-5-2-4

InChI key

JKWMSGQKBLHBQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anhydride can be obtained by heating boric acid at temperatures higher than 160oC.1

Application

Reducing boron anhydride in the presence of sodium yields amorphous boron. Whereas reducing in the presence of aluminum at high temperatures affords crystalline boron. 1

Packaging

Packaged in poly bottles

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinling Wang et al.
Analytical biochemistry, 385(1), 184-186 (2008-11-27)
Glassy carbon electrode modified with boron oxide nanoparticles supported on multiwall carbon nanotubes was obtained via a facile approach. The as-prepared modified electrode exhibits excellent electrocatalytic activity toward the redox of glucose in pH 7.0 phosphate buffer solution. The electrochemical
Xiaofeng Liu et al.
Chemical communications (Cambridge, England), (27)(27), 4073-4075 (2009-07-02)
A facile strategy based on the reaction of urea and boric acid is developed for the synthesis of toxicity-free BCNO nanocrystals containing carbon impurities and which exhibit efficient multi-color fluorescence under both single-photon and two-photon excitation.
V Simon et al.
Journal of materials science. Materials in medicine, 18(3), 507-512 (2007-03-06)
Quaternary Ag(2)O-CaO-Bi(2)O(3)-B(2)O(3) glasses and glassceramics are investigated with regard to release behaviour and local structure. The dissolution behaviour in water and physiological serum shows that the cations are released rapidly or gradually and points out a multi-step process, generally characterised
Pratik Koirala et al.
The journal of physical chemistry. A, 117(6), 1310-1318 (2012-11-03)
Using density functional theory based calculations, we have systematically studied the equilibrium geometries, relative stabilities, and electronic and magnetic properties of Fe and Mn atoms interacting with a varying number of BO(2) moieties. These clusters are found to exhibit hyperhalogen
T Raghavendra Rao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1116-1122 (2011-06-10)
Optical and physical properties of Ni2+ doped 20ZnO+xLi2O+(30-x)Na2O+50B2O3 (5≤x≤25) glasses are carried out at room temperature. Powder XRD pattern of all the glass samples confirms the amorphous nature. Several physical parameters are evaluated for all the glasses with respect to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service