Merck
  • Mixed alkali effect and optical properties of Ni2+ doped 20ZnO+xLi2O+(30-x)Na2O+50B2O3 glasses.
Mixed alkali effect and optical properties of Ni2+ doped 20ZnO+xLi2O+(30-x)Na2O+50B2O3 glasses.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-10)
T Raghavendra Rao, Ch Rama Krishna, Ch Venkata Reddy, U S Udayachandran Thampy, Y P Reddy, P S Rao, R V S S N Ravikumar
ABSTRACT
Optical and physical properties of Ni2+ doped 20ZnO+xLi2O+(30-x)Na2O+50B2O3 (5≤x≤25) glasses are carried out at room temperature. Powder XRD pattern of all the glass samples confirms the amorphous nature. Several physical parameters are evaluated for all the glasses with respect to the composition. The optical absorption spectra confirm the site symmetry of the Ni2+ doped glasses are near octahedral. Crystal field and inter-electronic repulsion parameters are also evaluated. It is interesting to observe that the optical band gap and Urbach energies exhibit the mixed alkali effect. The FT-IR spectral investigations of Ni2+ doped glasses exhibit characteristic vibrations of BO3 and BO4 units.
Product Description
Supelco
Boric anhydride, 99.98% trace metals basis
Sigma-Aldrich
Boric anhydride, granulated, ≥98.0% (T)
Sigma-Aldrich
Boric anhydride, 99.999% trace metals basis
Sigma-Aldrich
Boric anhydride, puriss. p.a., ≥98% (T)
Sigma-Aldrich
Sodium oxide, 80%