Skip to Content
Merck
All Photos(1)

Key Documents

18086

Sigma-Aldrich

1-Ethyl-3-methylimidazolium methyl sulfate

≥98.0% (HPLC)

Synonym(s):

[EMIM][MS]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4S
CAS Number:
Molecular Weight:
222.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

composition

carbon, 37.07-38.58%
nitrogen, 12.35-12.85% (as tested by elemental analysis)

impurities

≤0.2% water

SMILES string

COS([O-])(=O)=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.CH4O4S/c1-3-8-5-4-7(2)6-8;1-5-6(2,3)4/h4-6H,3H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

BXSDLSWVIAITRQ-UHFFFAOYSA-M

General description

1-Ethyl-3-methylimidazolium methyl sulfate [EMIM][MS] when added as an eluent modifier to the mobile phase results in better separation of the three positional isomers of benzoic acid by reversed-phase (RP) high-performance liquid chromatography (HPLC).

Application

[EMIM][MS] may be used as a solvent to synthesize gold nanoparticles via reduction of gold(III) chloride trihydrate [HAuCl4.3H2O] using glycerol as a reducing agent.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of ionic liquid as additive and the pH of the mobile phase on the retention factors of amino benzoic acids in RP-HPLC.
Zheng J, et al.
Journal of Chromatographic Science, 45(5), 256-262 (2007)
Retention factors and resolutions of amino benzoic acid isomers with some lonic liquids.
Zheng J, et al.
Biotechnology and Bioprocess Engineering: BBE, 11(6), 477-483 (2006)
Densities and viscosities of 1-ethyl-3-methylimidazolium n-alkyl sulfates
Costa, Anabela JL, et al.
Journal of Chemical and Engineering Data, 56.8, 3433-3441 (2011)
Strong anion effects on gold nanoparticle formation in ionic liquids.
Khare V, et al.
Journal of Materials Chemistry, 20(7), 1332-1339 (2010)
Microwave-assisted separation of ionic liquids from aqueous solution of ionic liquids
Ha, Sung Ho, Ngoc Lan Mai, and Yoon-Mo Koo
Journal of Chromatography A, 1217.49, 7638-7641 (2010)

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service