Skip to Content
Merck
All Photos(1)

Key Documents

39952

Sigma-Aldrich

1-Butyl-3-methylimidazolium acetate

≥95%

Synonym(s):

1-Butyl-3-methyl-1H-imidazol-3-ium acetate, 1-Methyl-3-butylimidazolium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N2O2
CAS Number:
Molecular Weight:
198.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

liquid

impurities

≤2.0% water

SMILES string

CC([O-])=O.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.C2H4O2/c1-3-4-5-10-7-6-9(2)8-10;1-2(3)4/h6-8H,3-5H2,1-2H3;1H3,(H,3,4)/q+1;/p-1

InChI key

BSKSXTBYXTZWFI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Butyl-3-methylimidazolium acetate is a 1-alkyl-3-methyl imidazolium based ionic liquid.

Application

1-Butyl-3-methylimidazolium acetate can be used as an additive to electrolytes for non-aqueous capillary electrophoresis (NACE) due to its air- and water-stability and miscibility with most of CE solvents.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility and density of carbon dioxide in different aqueous alkanolamine solutions blended with 1-butyl-3-methylimidazolium acetate ionic liquid at high pressure
Shojaeian, Abolfazl, and Ali Haghtalab
Journal of Molecular Liquids, 187, 218-225 (2013)
Flexural and impact properties of all-cellulose composite laminates
Huber, Tim, et al.
Composites Science and Technology, 88, 92-98 (2013)
Comparison of the thermal denaturing of human serum albumin in the presence of guanidine hydrochloride and 1-butyl-3-methylimidazolium ionic liquids
Heller, William T.
The Journal of Physical Chemistry B, 117.8, 2378-2383 (2013)
Solvent infusion processing of all-cellulose composite materials
Huber, Tim, et al.
Carbohydrate Polymers, 90.1, 730-733 (2012)
High pressure measurement and CPA equation of state for solubility of carbon dioxide and hydrogen sulfide in 1-butyl-3-methylimidazolium acetate
Haghtalab, Ali, and Alireza Kheiri
The Journal of Chemical Thermodynamics, 89, 41-50 (2015)

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service