Skip to Content
Merck
All Photos(1)

Key Documents

432628

Sigma-Aldrich

α-Terpineol

90%, technical grade

Synonym(s):

alpha-Terpineol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2325137
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical grade

Quality Level

Assay

90%

refractive index

n20/D 1.482 (lit.)

bp

217-218 °C (lit.)

mp

31-35 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC1=CC[C@H](CC1)C(C)(C)O

InChI

1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3

InChI key

WUOACPNHFRMFPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity in Melaleuca alternifolia (tea tree) essential oil.[1] The reaction rate constant of α-terpineol with the OH radical and ozone was found to be (1.9±0.5)×10-10cm3 molecule-1s-1 and (3.0±0.2)×10-16cm3 molecule-1s-1, respectively.[2]

Application

  • a-Terpineol, a natural monoterpene: A review of its biological properties: This review article explores the wide range of biological activities of a-terpineol, including antimicrobial, anti-inflammatory, and anticancer properties, providing a comprehensive overview of its therapeutic potential (C Khaleel, N Tabanca, G Buchbauer, 2018).
  • Production, properties, and applications of a-terpineol: Discusses the production methods, physical and chemical properties, and various industrial applications of a-terpineol, highlighting its importance in food and bioprocess technologies (A Sales et al., 2020).
  • Antifungal activity of citral, octanal and a-terpineol against Geotrichum citri-aurantii: This article examines the effectiveness of a-terpineol, along with other compounds, in inhibiting the growth of a fungal pathogen affecting citrus fruits, showcasing its potential as a natural fungicide (H Zhou et al., 2014).

Other Notes

Optical rotation varies with isomer mix.
remainder γ-isomer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annarica Calcabrini et al.
The Journal of investigative dermatology, 122(2), 349-360 (2004-03-11)
The search for innovative therapeutic approaches based on the use of new substances is gaining more interest in clinical oncology. In this in vitro study the potential anti-tumoral activity of tea tree oil, distilled from Melaleuca alternifolia, was analyzed against
Francesca Mondello et al.
BMC infectious diseases, 6, 158-158 (2006-11-07)
Recent investigations on the antifungal properties of essential oil of Melaleuca alternifolia Cheel (Tea Tree Oil, TTO) have been performed with reference to the treatment of vaginal candidiasis. However, there is a lack of in vivo data supporting in vitro
P H Hart et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 49(11), 619-626 (2000-12-29)
To evaluate potential antiinflammatory properties of tea tree oil, the essential oil steam distilled from the Australian native plant, Melaleuca alternifolia. The ability of tea tree oil to reduce the production in vitro of tumour necrosis factor-alpha (TNFalpha), interleukin (IL)-1beta
Jason E Ham et al.
Chemosphere, 83(3), 327-333 (2011-01-18)
Indoor environments are dynamic reactors where consumer products (such as cleaning agents, deodorants, and air fresheners) emit volatile organic compounds (VOCs) that can subsequently interact with indoor oxidants such as ozone (O(3)), hydroxyl radicals, and nitrate radicals. Typically, consumer products
Hanne Halkinrud Thoen et al.
The Journal of comparative neurology, 528(7), 1079-1094 (2019-10-18)
Mantis shrimps (Stomatopoda) possess in common with other crustaceans, and with Hexapoda, specific neuroanatomical attributes of the protocerebrum, the most anterior part of the arthropod brain. These attributes include assemblages of interconnected centers called the central body complex and in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service