Skip to Content
Merck
All Photos(1)

Key Documents

147982

Sigma-Aldrich

(S)-(+)-2-Octanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719323
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]20/D +9.5°, neat

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.426 (lit.)

bp

175 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[C@H](C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3/t8-/m0/s1

InChI key

SJWFXCIHNDVPSH-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-2-Octanol can be used:
  • To prepare the solution of 5-(benzyloxy)-isophthalic acid derivative, which is used as a 2D chiral self-assembly system comprising pyrolytic graphite.
  • As a chiral template in the study of enantioselective glycidol esterification.
  • As a starting material for the preparation of (+)-(S)-2-octyI tosylate, an intermediate used to prepare (−)-(R)-2-halo and azido octanes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convenient preparation of optically active 2-halooctanes and related compounds
San Filippo Jr J and Romano LJ
The Journal of Organic Chemistry, 40(10), 1514-1515 (1975)
Paul Masset et al.
Cell, 182(1), 112-126 (2020-06-07)
Every decision we make is accompanied by a sense of confidence about its likely outcome. This sense informs subsequent behavior, such as investing more-whether time, effort, or money-when reward is more certain. A neural representation of confidence should originate from
Xiao-Yang Ou et al.
Journal of biotechnology, 299, 37-43 (2019-05-03)
Highly efficient asymmetric reduction of 2-octanone to (R)-2-octanol catalyzed by immobilized Acetobacter sp. CCTCC M209061 cells was achieved in a biphasic system. Bioreduction conducted in aqueous single phase buffer was limited due to poor solubility and toxicity towards cells cause
Development of simulation approach for two-dimensional chiral molecular self-assembly driven by hydrogen bond at the liquid/solid interface
Qin Y, et al.
Surface Science, 663, 71-80 (2017)
Effect of imprinting sol-gel immobilized lipase with chiral template substrates in esterification of (R)-(+)-and (S)-(-)-glycidol.
Furukawa S, et al.
Journal of Molecular Catalysis. B, Enzymatic, 17(1), 23-28 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service