Skip to Content
Merck
All Photos(1)

Key Documents

74858

Sigma-Aldrich

(±)-2-Octanol

ReagentPlus®, ≥99.5% (GC)

Synonym(s):

Hexyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719325
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
grade:
reagent

grade

reagent

product line

ReagentPlus®

Assay

≥99.5% (GC)

form

liquid

dilution

(for general lab use)

refractive index

n20/D 1.426

density

0.819 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCC(C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3

InChI key

SJWFXCIHNDVPSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Yamamoto et al.
Applied microbiology and biotechnology, 97(18), 8087-8096 (2013-01-01)
A novel enantioselective alcohol dehydrogenase, (R)-2-octanol dehydrogenase (PfODH), was discovered among methylotrophic microorganisms. The enzyme was purified from Pichia finlandica and characterized. The molecular mass of the enzyme was estimated to be 83,000 and 30,000 by gel filtration and sodium
P G Aitken et al.
Journal of neurophysiology, 80(3), 1514-1521 (1998-09-24)
Neuron membrane changes and ion redistribution during normoxic spreading depression (SD) induced, for example, by potassium injection, closely resemble those that occur during hypoxic SD-like depolarization (HSD) induced by oxygen withdrawal, but the degree to which the two phenomena are
Er-na Xun et al.
Applied biochemistry and biotechnology, 168(3), 697-707 (2012-08-01)
The lipase from Pseudomonas fluorescens (Lipase AK, AKL) was immobilized onto the magnetic Fe(3)O(4) nanoparticles via hydrophobic interaction. Enzyme loading and immobilization yield were determined as 21.4±0.5 mg/g and 49.2±1.8 %, respectively. The immobilized AKL was successfully used for resolution
Deracemization of secondary alcohols through a concurrent tandem biocatalytic oxidation and reduction.
Constance V Voss et al.
Angewandte Chemie (International ed. in English), 47(4), 741-745 (2007-12-12)
M Martinelle et al.
Biochimica et biophysica acta, 1251(2), 191-197 (1995-09-06)
The acyl transfer reactions catalysed by Candida antartica lipase B in organic media followed a bi-bi ping-pong mechanism, with competitive substrate inhibition by the alcohols used as acyl acceptors. The effect of organic solvents on Vm and Km was investigated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service