Skip to Content
Merck
All Photos(1)

Key Documents

340332

Sigma-Aldrich

(S)-(+)-2-Heptanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH3
CAS Number:
Molecular Weight:
116.20
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

vapor density

4 (vs air)

vapor pressure

1 mmHg ( 15 °C)

Assay

98%

form

liquid

optical activity

[α]24/D +10°, neat

optical purity

ee: 97% (GLC)

refractive index

n20/D 1.421 (lit.)

bp

149-150 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)

SMILES string

CCCCC[C@H](C)O

InChI

1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3/t7-/m0/s1

InChI key

CETWDUZRCINIHU-ZETCQYMHSA-N

Application

(S)-(+)-2-Heptanol can be used as a starting material to synthesize:
  •  (S)-(+)-2-benzoyloxyheptane by treating with benzoyl anhydride and Et3N in the presence of DMAP.       
  • (S)-2-(p-toluenesulfonyl)heptane by reacting with p-toluenesulfonyl chloride.      
  •   (+)-2-Heptyl acetate by treating with acetyl chloride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective multidimensional gas chromatography of some secondary alcohols and their acetates from banana
Schubert, V, et al.
Zeitschrift fur Naturforschung B, 46(1-2), 33-36 (1991)
Christiane Kiske et al.
Journal of agricultural and food chemistry, 64(45), 8563-8571 (2016-11-04)
The absolute configurations of chiral β-mercaptoalkanones were previously assigned on the basis of the 1H NMR anisotropy method using (S)-2-methoxy-2-(1-naphthyl)propionic acid ((S)-MαNP) as the chiral auxiliary. This study presents a reinvestigation of the configurations of 4-mercapto-2-pentanone 1, 4-mercapto-2-heptanone 2, and
A study on the stereochemistry of direct conversion of polyamides to hydroxyesters using monomeric secondary chiral amines as a model compound
Kamimura A, et al.
Polymer Degradation and Stability, 160, 162-167 (2019)

Articles

Chiral Alcohols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service