Skip to Content
Merck
All Photos(1)

Key Documents

140406

Sigma-Aldrich

3,5-Ditert-butyl-4-hydroxybenzaldehyde

99%

Synonym(s):

2,6-Di-tert-Butyl-4-formylphenol, 3,5-Di-tert-butyl-4-hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

solubility

ethanol: soluble 50 mg/mL, clear to slightly hazy (yellow to orange to light red-brown)

functional group

aldehyde

SMILES string

CC(C)(C)c1cc(C=O)cc(c1O)C(C)(C)C

InChI

1S/C15H22O2/c1-14(2,3)11-7-10(9-16)8-12(13(11)17)15(4,5)6/h7-9,17H,1-6H3

InChI key

DOZRDZLFLOODMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Di-tert-butyl-4-hydroxybenzaldehyde can be used as a starting material to synthesize:

  • Biodegradable polyazomethine hydrogels by reacting with melamine via oxidative polymerization reaction.
  • 4-(1H-benzimidazol-2-yl)-2,6-di-tert butyl-phenol by treating with 1,2-phenylenediamine in ethanol.
  • 2-{(3,5-di-tert-butyl-4-hydroxyphenyl)methylene}-4-cyclopentene-1,3-dione (TX-1123) as a potent protein tyrosine kinase inhibitor.
  • Polyazomethine and ascorbic acid-based selective and sensitive fluorescent chemosensor for the detection of Al and Fe metal ions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melamine-based poly (azomethine) hydrogels: Mechanical, biodegradability, drug loading and antibacterial properties
Kamaci M and Kaya I
European Polymer Journal, 108, 107-115 (2018)
Highly stabilized phenoxyl radicals with hydrogen-bonding capability.
Xie C and Lahti PM.
Tetrahedron Letters, 40(23), 4305-4308 (1999)
A dual responsive colorimetric sensor based on polyazomethine and ascorbic acid for the detection of Al (III) and Fe (II) ions
Kamaci UD, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 254, 119650-119650 (2021)
Highly stabilized phenoxyl radicals with hydrogen-bonding capability
Xie Chunping and Lahti PM
Tetrahedron Letters, 40(23), 4305-4308 (1999)
TX-1123: an antitumor 2-hydroxyarylidene-4-cyclopentene-1, 3-dione as a protein tyrosine kinase inhibitor having low mitochondrial toxicity
Hori H, et al.
Bioorganic & Medicinal Chemistry Letters, 10(10), 3257-3265 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service