Skip to Content
Merck
All Photos(1)

Documents

137731

Sigma-Aldrich

2,4-Di-tert-butylphenol

99%

Synonym(s):

2,4-Bis(1,1-dimethylethyl)phenol, 2,4-Bis(tert-butyl)phenol, 2,4-Di-tert-butylhydroxybenzene, 2,4-Ditert-butylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3OH
CAS Number:
Molecular Weight:
206.32
Beilstein:
1910383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 84.5 °C)

Quality Level

Assay

99%

form

solid

bp

265 °C (lit.)

mp

53-56 °C (lit.)

solubility

water: soluble 0.033 g/L at 25 °C

SMILES string

CC(C)(C)c1ccc(O)c(c1)C(C)(C)C

InChI

1S/C14H22O/c1-13(2,3)10-7-8-12(15)11(9-10)14(4,5)6/h7-9,15H,1-6H3

InChI key

ICKWICRCANNIBI-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,4-Di-tert-butylphenol is isolated from the dried body of Scolopendra and possess antioxidative activity.

Application

2,4-Di-tert-butylphenol was used to study its alkylation reaction with cinnamyl alcohol in the presence of Al-substituted disordered hexagonal molecular silica catalysts.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sherry Cox et al.
PeerJ, 5, e3476-e3476 (2017-07-05)
Propofol is an intravenous anesthetic commonly used due to its favorable pharmacokinetic and pharmacodynamic profile. There are discrepancies in the literature about the most appropriate sample for determining propofol concentrations. Although plasma has been used for determining propofol concentrations, whole
Textural mesoporosity and the catalytic activity of mesoporous molecular sieves with wormhole framework structures.
Pauly TR, et al.
Journal of the American Chemical Society, 121(38), 8835-8842 (1999)
Jae Kwak et al.
PloS one, 11(3), e0151474-e0151474 (2016-03-12)
Male house mice produce large quantities of major urinary proteins (MUPs), which function to bind and transport volatile pheromones, though they may also function as scavengers that bind and excrete toxic compounds ('toxic waste hypothesis'). In this study, we demonstrate
Verónica Ramírez et al.
Frontiers in microbiology, 11, 741-741 (2020-05-20)
Volatile organic compounds (VOCs) produced by rhizobacteria have been proven to stimulate plant growth during germination and seedling stages. However, the modulating effect of bacterial volatiles on the germination of seeds subjected to heavy metal stress is scarcely studied. In
Noemí Dorival-García et al.
Journal of chromatography. A, 1513, 69-77 (2017-07-26)
Despite their advantages, there is concern that single-use systems used in biopharmaceutical manufacture might release potentially toxic substances during standard unit operations that negatively impact cell growth. Characterization of the extractables profile for single-use systems is necessary to know which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service