Skip to Content
Merck
All Photos(1)

Key Documents

140414

Sigma-Aldrich

3,5-Di-tert-butyl-2-hydroxybenzaldehyde

99%

Synonym(s):

3,5-Di-tert-butylsalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2[C(CH3)3]2CHO
CAS Number:
Molecular Weight:
234.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder or crystals

mp

59-61 °C (lit.)

functional group

aldehyde

SMILES string

CC(C)(C)c1cc(C=O)c(O)c(c1)C(C)(C)C

InChI

1S/C15H22O2/c1-14(2,3)11-7-10(9-16)13(17)12(8-11)15(4,5)6/h7-9,17H,1-6H3

InChI key

RRIQVLZDOZPJTH-UHFFFAOYSA-N

General description

3,5-Di-tert-butyl-2-hydroxybenzaldehyde undergoes condensation reaction with
  • methyl-2-{N-(2′-aminoethane)}-amino-1-cyclopentenedithiocarboxylate to yield Schiff base ligand
  • N,N-diethyl-2-methyl-1,4-phenylenediamine during the synthesis of copper(II) and cobalt(II) complexes of salicylaldimine

Application

3,5-Di-tert-butyl-2-hydroxybenzaldehyde was used in the synthesis of
  • Mn(III)-salen complex and its diamino precursor 5,6-diamino-5,6-dideoxy-1,2-O-isopropylidene-3-O-methyl-β-L-idofuranose
  • chiral Schiff base ligand for an enantioselective copper-catalyzed addition of phenylacetylene to imines
  • chiral oxazolidine ligand for the enantioselective addition of diethylzinc to aldehydes
  • tin Schiff base complexes with histidine analogues

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mild and efficient synthesis of 5, 6-diamino-5, 6-dideoxy-1, 2-O-isopropylidene-3- O -methyl-?-l-idofuranose: precursor of the first carbohydrate-derived chiral Mn (III)-salen complex.
Yan S and Klemm D.
Tetrahedron, 58(50), 10065-10071 (2002)
Ariadna Garza-Ortiz et al.
Bioinorganic chemistry and applications, 2013, 502713-502713 (2013-07-19)
Five novel tin Schiff base complexes with histidine analogues (derived from the condensation reaction between L-histidine and 3,5-di-tert-butyl-2-hydroxybenzaldehyde) have been synthesized and characterized. Characterization has been completed by IR and high-resolution mass spectroscopy, 1D and 2D solution NMR ((1)H, (13)C  and
Co (II) and Cu (II) Schiff base complexes of bis (N-(4-diethylamino-2-methylphenyl)-3, 5-di-tert-butylsalicylaldimine): Electrochemical and X-ray structural study.
Ulusoy M, et al.
Structural Chemistry, 19(5), 749-755 (2008)
Tetrahedron Asymmetry, 18, 1124-1124 (2007)
Tetrahedron Asymmetry, 18, 377-377 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service