Skip to Content
Merck
All Photos(2)

Documents

135178

Sigma-Aldrich

Cuminaldehyde

98%

Synonym(s):

4-Isopropylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4CHO
CAS Number:
Molecular Weight:
148.20
Beilstein:
636547
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

235-236 °C (lit.)

solubility

H2O: insoluble
alcohol: soluble
diethyl ether: soluble

density

0.981 g/mL at 20 °C
0.977 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(cc1)C(C)C

InChI

1S/C10H12O/c1-8(2)10-5-3-9(7-11)4-6-10/h3-8H,1-2H3

InChI key

WTWBUQJHJGUZCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cuminaldehyde has been used to study larvicidal and adulticidal toxicity of monoterpenes against Culex pipiens. It has been used in evaluation of chemical composition, antimicrobial and antioxidant activities of essential oil and various extracts of Eucalyptus gilii.

Biochem/physiol Actions

Cuminaldehyde increases the insulin secretion in streptozotocin-induced diabetic rats. It suppresses melanin formation in cultured murine B16-F10 melanoma cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

LARVICIDAL, ADULTICIDAL AND GROWTH INHIBITORY EFFECTS OF MONOTERPENES ON CULEX PIPIENS L.(DIPTERA: CULICIDAE).
Zahran HE-DM, et al.
Journal of Agricultural Research Kafer El-Sheikh Univ, 36(4), 385-341 (2010)
Dorsaf Ben Hassine et al.
Molecules (Basel, Switzerland), 17(8), 9540-9558 (2012-08-11)
In this study, essential oil and various extracts (hexane, petroleum ether, acetone, ethanol, methanol and water) of Eucalyptus gilii were screened for their chemical composition, antimicrobial and antioxidant activities. The essential oil chemical composition was analyzed by gas chromatography-mass spectrometry
T P Krishnakantha et al.
Indian journal of biochemistry & biophysics, 30(2), 133-134 (1993-04-01)
Effect of spice principles on scavenging of superoxide anion has been investigated. The superoxide anions, as measured by nitrobluetetrazolium (NBT) reduction in xanthine-xanthine oxidase system, were inhibited by superoxide dismutase, spice principles eugenol (cloves) and cuminaldehyde (cumin), antioxidants, butylated hydroxy
M Désage et al.
Journal of chromatography. B, Biomedical applications, 678(2), 205-210 (1996-04-12)
The flavours contained in a mammalian mother's milk can exert a marked influence on her offspring's proximate suckling behaviour and later preferences. The aim of this study was to establish a reliable analytical procedure to characterise the mammary transfer of
Hoi-Seon Lee
Journal of agricultural and food chemistry, 53(7), 2446-2450 (2005-03-31)
The inhibitory activity of Cuminum cyminum seed-isolated component was evaluated against lens aldose reductase and alpha-glucosidase isolated from Sprague-Dawley male rats and compared to that of 11 commercially available components derived from C. cyminum seed oil, as well as quercitrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service