Skip to Content
Merck
All Photos(1)

Documents

T35602

Sigma-Aldrich

p-Tolualdehyde

97%

Synonym(s):

4-Methylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CHO
CAS Number:
Molecular Weight:
120.15
Beilstein:
385772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.545 (lit.)

bp

204-205 °C (lit.)
82-85 °C/11 mmHg (lit.)

solubility

water: soluble 0.25 g/L at 25 °C

density

1.019 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(C)cc1

InChI

1S/C8H8O/c1-7-2-4-8(6-9)5-3-7/h2-6H,1H3

InChI key

FXLOVSHXALFLKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Tolualdehyde (4-Methylbenzaldehyde) is an aromatic aldehyde. It has been generated as major oxygenated product during the UV light irradiated oxygenation of p-xylene, via photoinduced electron transfer mechanism. It undergoes condensation reaction with diiron μ-ethylidyne complex to afford μ-vinylcarbyne complex (92% yield).

Application

p-Tolualdehyde may be employed as growth substrate in the culture medium of the strains of Pseudomonas putida.
p-Tolualdehyde may be used in the preparation of (E)-3-(4-methyl-phen-yl)-1-(1,3-thia-zol-2-yl)prop-2-en-1-one. It may be used in the preparation of (Z)-trisubstituted allylic alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ohkubo et al.
Organic letters, 2(23), 3647-3650 (2000-11-14)
The 100% selective oxygenation of p-xylene to p-tolualdehyde is initiated by photoinduced electron transfer from p-xylene to the singlet excited state of 10-methyl-9-phenylacridinium ion under visible light irradiation, yielding p-tolualdehyde exclusively as the final oxygenated product. The reason for the
Young K Chen et al.
Journal of the American Chemical Society, 126(12), 3702-3703 (2004-03-25)
In this Communication, we outline a new one-pot, multicomponent coupling reaction that allows easy access to (Z)-trisubstituted allylic alcohols. Our strategy is based on E to Z isomerization of the 1-bromo-1-dialkylvinylborane upon reaction with dialkylzinc reagents, and subsequent transmetalation to
M J Worsey et al.
Journal of bacteriology, 124(1), 7-13 (1975-10-01)
Pseudomonas putida (arvilla) mt-2 carries genes for the catabolism of toluene, m-xylene, and p-xylene on a transmissible plasmid, TOL. These compounds are degraded by oxidation of one of the methyl substituents via the corresponding alcohols and aldehydes to benzoate and
Synthesis of cationic diiron μ-vinylcarbyne complexes.
Casey CP, et al.
Polyhedron, 7(10), 881-992 (1988)
Annamalai Palaniappan et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 6), o1875-o1875 (2012-06-22)
In the title chalcone, C(13)H(11)NOS, derived from the condensation of p-tolualdehyde and 1-(1,3-thia-zol-2-yl)ethanone, the olefine group has a trans configuration. No classical hydrogen bonding is present in the crystal structure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service