132071
1,3,5-Triethylbenzene
≥97%
Synonym(s):
1,3,5-Triethylbenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H3(C2H5)3
CAS Number:
Molecular Weight:
162.27
Beilstein:
2039481
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥97%
form
liquid
refractive index
n20/D 1.495 (lit.)
bp
215 °C (lit.)
density
0.862 g/mL at 25 °C (lit.)
SMILES string
CCc1cc(CC)cc(CC)c1
InChI
1S/C12H18/c1-4-10-7-11(5-2)9-12(6-3)8-10/h7-9H,4-6H2,1-3H3
InChI key
WJYMPXJVHNDZHD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
168.8 °F - closed cup
Flash Point(C)
76 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Xing Wang et al.
Beilstein journal of organic chemistry, 8, 1-10 (2012-03-17)
1,3,5-triethylbenzenes have been widely used as supramolecular templates to organize molecular-recognition elements. It is believed that the steric-gearing effect of the 1,3,5-triethylbenzene template directs the binding elements toward the same face of the central ring, hence increasing the binding affinity.
Gou Higashihara et al.
Dalton transactions (Cambridge, England : 2003), (14)(14), 1888-1898 (2008-03-29)
A series of di- and trinucleating ligands with a 1,3,5-triethylbenzene core connected to N,N-bidentate tethers was synthesized. The ligands readily reacted with monuclear Rh and Pd precursors to give the corresponding di- and trinuclear complexes, which were characterized by using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service