T19607
1,2,4,5-Tetramethylbenzene
98%
Synonym(s):
Durene, 1,2,4,5-Tetramethylbenzene
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C6H2(CH3)4
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903393
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39011404
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
4.6 (vs air)
vapor pressure
160 mmHg ( 140 °C)
Assay
98%
mp
76-80 °C (lit.)
density
0.838 g/mL at 25 °C (lit.)
SMILES string
Cc1cc(C)c(C)cc1C
InChI
1S/C10H14/c1-7-5-9(3)10(4)6-8(7)2/h5-6H,1-4H3
InChI key
SQNZJJAZBFDUTD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 1
Flash Point(F)
165.2 °F - closed cup
Flash Point(C)
74 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Joaquín Ortego et al.
Molecular ecology, 24(23), 5796-5812 (2015-10-18)
Conservation plans can be greatly improved when information on the evolutionary and demographic consequences of habitat fragmentation is available for several codistributed species. Here, we study spatial patterns of phenotypic and genetic variation among five grasshopper species that are codistributed
Elin Rönnberg et al.
Immunology, 143(2), 155-163 (2014-04-03)
Staphylococcus aureus is a major pathogen that can cause a broad spectrum of serious infections including skin infections, pneumonia and sepsis. Peritoneal mast cells have been implicated in the host response towards various bacterial insults and to provide mechanistic insight
S David et al.
Journal of inorganic biochemistry, 28(2-3), 125-135 (1986-10-01)
Solution equilibria are presented for in situ reactions of the type FeII(porp) + B----FeII(porp)B, where porp represents the dianion of some durene-capped porphyrins with variable length linking methylene straps, and B is 1-methyl, 1,2-dimethyl, or 1,5-dicyclohexylimidazole. Increasing distortion of the
D Reichert et al.
Solid state nuclear magnetic resonance, 13(3), 137-148 (1999-02-19)
One- (1-D) and two-dimensional (2-D) carbon-13 NMR exchange measurements in powder samples of isotopically normal durene under magic angle spinning (MAS) are reported. The experiments include rotor synchronized 2-D exchange (RS2DE), 1-D magnetization transfer (MT) and time reverse ODESSA (tr-ODESSA).
Yuan Shen et al.
Plant biotechnology journal, 13(6), 811-820 (2015-01-13)
Chromatin modification and remodelling are the basis for epigenetic regulation of gene expression. LEAFY COTYLEDON 1 (LEC1), LEAFY COTYLEDON 2 (LEC2), ABSCISIC ACID-INSENSITIVE 3 (ABI3) and FUSCA3 (FUS3) are key regulators of embryo development and are repressed after seed maturation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service