Skip to Content
Merck
All Photos(1)

Key Documents

D91004

Sigma-Aldrich

1,4-Diethylbenzene

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(C2H5)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903396
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

0.99 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

806 °F

refractive index

n20/D 1.495 (lit.)

bp

184 °C (lit.)

mp

−43 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(CC)cc1

InChI

1S/C10H14/c1-3-9-5-7-10(4-2)8-6-9/h5-8H,3-4H2,1-2H3

InChI key

DSNHSQKRULAAEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F

Flash Point(C)

56 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Gagnaire et al.
Journal of applied toxicology : JAT, 12(5), 335-342 (1992-10-01)
Motor and sensory conduction velocities (MCV and SCV), amplitude of the sensory action potential (ASAP) of the tail nerve and parameters of brainstem auditory evoked potentials (BAEP) were studied in male Sprague-Dawley rats after prolonged inhalation exposure to a commercial
I Linhart et al.
Journal of chromatography, 530(2), 283-294 (1990-09-14)
The metabolism of 1,4-diethenylbenzene in the rat was followed by gas chromatographic-mass spectrometric analysis of urine using three different derivatization procedures: (i) methylation-acetylation; (ii) methylation-trimethylsilylation; (iii) methylation followed by conversion into trimethylsilyloximes. Fifteen metabolites were found in the urine of
Consuelo Pizarro et al.
Journal of chromatography. A, 1167(2), 202-209 (2007-09-14)
The binary diffusion of 1,2-diethylbenzene, 1,4-diethylbenzene, 5-tert-butyl-m-xylene and phenylacetylene at infinite dilution in supercritical carbon dioxide were measured between 15.0 and 35.0 MPa and in the temperature range of 313.16 to 333.16K by the Taylor-Aris chromatographic method. The effect of
F Gagnaire et al.
Journal of applied toxicology : JAT, 10(2), 105-112 (1990-04-01)
The commercial isomer mixture of diethylbenzene (DEB mixture), 1,2-diethylbenzene (1,2-DEB), 1,3-diethylbenzene (1,3-DEB) and 1,4-diethylbenzene (1,4-DEB) were administered orally to male Sprague-Dawley rats. The experimental rats and the appropriate controls were examined electrophysiologically for motor and sensory conduction velocities (MCV and
F Gagnaire et al.
Journal of applied toxicology : JAT, 12(5), 343-350 (1992-10-01)
Male Sprague-Dawley rats were treated either with 1,2-diethylbenzene (1,2-DEB) or its putative active metabolite, 1,2-diacetylbenzene (1,2-DAB). Experimental rats and appropriate controls were examined electrophysiologically for brainstem auditory evoked potentials (BAEP). Oral administration of 1,2-DEB (75 or 100 mg kg-1 once

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service