Skip to Content
Merck
All Photos(1)

Documents

129658

Sigma-Aldrich

m-Anisaldehyde

97%

Synonym(s):

3-Methoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
Beilstein:
606013
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.553 (lit.)

bp

143 °C/50 mmHg (lit.)

density

1.117 g/mL at 20 °C (lit.)

SMILES string

[H]C(=O)c1cccc(OC)c1

InChI

1S/C8H8O2/c1-10-8-4-2-3-7(5-8)6-9/h2-6H,1H3

InChI key

WMPDAIZRQDCGFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Anisaldehyde has significant antifungal activity against Candida sp., including the azole-resistant strains.

m-Anisaldehyde is a class of benzaldehyde, used as a potent antifungal agent and as the starting material for complex aromatic compounds.

Application

m-Anisaldehyde was used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It was also used as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Morse et al.
Cancer letters, 97(2), 255-261 (1995-11-06)
As part of a routine screening assay, benzaldehyde was found to inhibit 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism. Consequently, the effects of benzaldehyde and several structurally related compounds on NNK metabolism were examined in murine hepatic and pulmonary microsomes. All test compounds inhibited
Structural and spectroscopic (UV--Vis, IR, Raman, and NMR) characteristics of anisaldehydes that are flavoring food additives: A density functional study in comparison with experiments
Altun, et al.
Journal of Molecular Structure, 1128, 590-605 (2017)
Retention of sparingly soluble volatile compounds during the freeze drying of model solutions
Smyrl,et al.
Journal of Food Process Engineering, 2, 151-170 (1978)
Eliot P Botosoa et al.
Journal of chromatography. A, 1216(42), 7043-7048 (2009-09-15)
Quantitative isotopic (13)C NMR at natural abundance has been used to determine the site-by-site (13)C/(12)C ratios in vanillin and a number of related compounds eluted from silica gel chromatography columns under similar conditions. Head-to-tail isotope fractionation is observed in all
Sheikh Shreaz et al.
Microbial pathogenesis, 51(4), 277-284 (2011-06-15)
Attention has been drawn to evaluate the antifungal activity of p-anisaldehyde (1), o-anisaldehyde (2) and m-anisaldehyde (3). To put forward this approach, antifungal activity has been assessed in thirty six fluconazole-sensitive and eleven fluconazole-resistant Candida isolates. Growth and sensitivity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service