Skip to Content
Merck
All Photos(1)

Documents

97063

Supelco

p-Anisaldehyde

analytical standard

Synonym(s):

4-Methoxybenzaldehyde, Aubépine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
Beilstein:
471382
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.7 (vs air)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.573 (lit.)
n20/D 1.573

bp

248 °C (lit.)

mp

−1 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)c1ccc(OC)cc1

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

ZRSNZINYAWTAHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-anisaldehyde is an aromatic aldehyde commonly found in anise seed oil. It shows acaricidal activity and is primarily used, as a lead compound for the development of new agents for the selective control of house dust mites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

may discolor to yellow on storage

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p-Anisaldehyde: acaricidal component of Pimpinella anisum seed oil against the house dust mites Dermatophagoides farinae and Dermatophagoides pteronyssinus.
Lee S-H
Planta Medica, 70(03), 279-281 (2004)
Aromatic aldehydes as substrates for yeast and yeast alcohol dehydrogenase.
A Long et al.
Biotechnology and bioengineering, 33(5), 657-660 (1989-01-25)
Wenxin Gu et al.
Organic letters, 5(4), 415-418 (2003-02-14)
[reaction: see text] Two new 4-methoxybenzaldehyde backbone linker resins were developed for the solid-phase synthesis of peptides. The linkers are very stable during the cleavage of common protecting groups for amines (Fmoc, Boc) and carboxylic acids (Me, All, tBu) in
Israt Jahan et al.
Acta pharmaceutica (Zagreb, Croatia), 60(2), 207-215 (2010-12-08)
In this study, five compounds, lupeol (1), epilupeol (2), β-sitosterol (3), stigmasterol (4) and p-methoxybenzaldehyde (5) were isolated from the petroleum ether and dichloromethane fractions of a methanolic extract of the stem bark of Delonix regia. Antimicrobial screening of the
Guozhu Zhang et al.
Journal of the American Chemical Society, 130(38), 12598-12599 (2008-08-30)
A novel approach to generate Au-containing all-carbon 1,3-dipoles is developed via an unprecedented migration-fragmentation of ketals/acetals. These in situ generated dipoles undergo rapid [3+2] cycloaddition under mild conditions with various enones/enals, electron-rich aromatic aldehydes, and N-benzylindole, leading to flexible formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service