Merck
  • Synthesis and Catalytic Performance of Hierarchically Porous MIL-100(Fe)@polyHIPE Hybrid Membranes.
Synthesis and Catalytic Performance of Hierarchically Porous MIL-100(Fe)@polyHIPE Hybrid Membranes.
Macromolecular rapid communications (2015-07-15)
Sebastijan Kovačič, Matjaž Mazaj, Marjan Ješelnik, David Pahovnik, Ema Žagar, Christian Slugovc, Nataša Zabukovec Logar
ABSTRACT
Metal-organic frameworks (MOFs) nanoparticles in combination with a nonionic surfactant (Pluronic L-121) are used to stabilize dicyclopentadiene (DCPD)-in-water high internal phase emulsions (HIPEs). The resulting HIPEs containing the MIL-100(Fe) nanoparticles (MIL: Materials of Institut Lavoisier) at the interface between the oil- and the water-phases are then cured, and 100 μm thick, fully open, hierarchically porous hybrid membranes are obtained. The properties of the MIL-100(Fe)@pDCPD polyHIPE membranes are characterized and it is found that up to 14 wt% of the MIL-100(Fe) nanoparticles are incorporated in the hybrid material resulting in an increase of the microporosity up to 130 m(2) g(-1). Hybrid membranes show an appealing catalytic activity in Friedel-Crafts alkylation in a batch mode as well as in a flow-through mode, thereby demonstrating the preserved accessibility of Lewis acidic sites in the MOF nanostructures.
Product Description
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Tricyclohexylphosphine solution, 20 wt. % in toluene
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
Tricyclohexylphosphine