Skip to Content
Merck
All Photos(1)

Key Documents

1633506

USP

Sulfanilic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
908765
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mesalazine, sulfadimethoxine, sulfadiazine

manufacturer/tradename

USP

mp

>300 °C (lit.)

solubility

water: soluble 12.51 g/L at 20 °C

density

1.4862 g/cm3 at 20 °C (OECD Test Guideline 109)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfanilic acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Silver Sulfadiazine
  • Sulfamethoxazole
  • Sulfamethoxazole and Trimethoprim Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Luís Belo et al.
PloS one, 9(6), e98467-e98467 (2014-06-06)
Bilirubin has potential antioxidant and anti-inflammatory properties. The UGT1A1*28 polymorphism (TA repeats in the promoter region) is a major determinant of bilirubin levels and recent evidence suggests that raised adiposity may also be a contributing factor. We aimed to study
Aminul Islam et al.
Journal of AOAC International, 98(1), 165-175 (2015-04-11)
Amberlite® XAD-16 was functionalized with p-aminobenzene sulfonic acid via an azo spacer in order to prepare a new chelating resin, which was then characterized by water regain value, hydrogen ion capacity, elemental analyses, and IR spectral and thermal studies. The
Virginia A Pereira et al.
Malaria journal, 14, 30-30 (2015-01-30)
Cytokines play an important role in human immune responses to malaria and variation in their production may influence the course of infection and determine the outcome of the disease. The differential production of cytokines has been linked to single nucleotide
Xiaojun Zhou et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11834-11842 (2014-06-25)
This paper presents a facile method for the fabrication of uniform hollow mesoporous silica nanoparticles (HMSNs) with tunable shell thickness and pore size. In this method, a series of amphiphilic block copolymers of polystyrene-b-poly (acrylic acid) (PS-b-PAA) with different hydrophobic
Kraingkrai Ponhong et al.
Talanta, 133, 71-76 (2014-12-02)
A novel four-channel simultaneous injection effective mixing flow analysis (SIEMA) system has been assembled for successive determination of bilirubin and creatinine in urinary samples. The chemical variables and physical parameters in the flow system were optimized for the enhancement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service