Merck
  • Body fat percentage is a major determinant of total bilirubin independently of UGT1A1*28 polymorphism in young obese.
Body fat percentage is a major determinant of total bilirubin independently of UGT1A1*28 polymorphism in young obese.
PloS one (2014-06-06)
Luís Belo, Henrique Nascimento, Michaela Kohlova, Elsa Bronze-da-Rocha, João Fernandes, Elísio Costa, Cristina Catarino, Luísa Aires, Helena Ferreira Mansilha, Petronila Rocha-Pereira, Alexandre Quintanilha, Carla Rêgo, Alice Santos-Silva
ABSTRACT
Bilirubin has potential antioxidant and anti-inflammatory properties. The UGT1A1*28 polymorphism (TA repeats in the promoter region) is a major determinant of bilirubin levels and recent evidence suggests that raised adiposity may also be a contributing factor. We aimed to study the interaction between UGT1A1 polymorphism, hematological and anthropometric variables with total bilirubin levels in young individuals. 350 obese (mean age of 11.6 years; 52% females) and 79 controls (mean age of 10.5 years; 59% females) were included. Total bilirubin and C-reactive protein (CRP) plasma levels, hemogram, anthropometric data and UGT1A1 polymorphism were determined. In a subgroup of 74 obese and 40 controls body composition was analyzed by dual-energy X-ray absorptiometry. The UGT1A1 genotype frequencies were 49.9%, 42.7% and 7.5% for 6/6, 6/7 and 7/7 genotypes, respectively. Patients with 7/7 genotype presented the highest total bilirubin levels, followed by 6/7 and 6/6 genotypes. Compared to controls, obese patients presented higher erythrocyte count, hematocrit, hemoglobin and CRP levels, but no differences in bilirubin or in UGT1A1 genotype distribution. Body fat percentage was inversely correlated with bilirubin in obese patients but not in controls. This inverse association was observed either in 6/7 or 6/6 genotype obese patients. UGT1A1 polymorphism and body fat percentage were the main factors affecting bilirubin levels within obese patients (linear regression analysis). In obese children and adolescents, body fat composition and UGT1A1 polymorphism are independent determinants of total bilirubin levels. Obese individuals with 6/6 UGT1A1 genotype and higher body fat mass may benefit from a closer clinical follow-up.
Product Description
USP
Sulfanilic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Supelco
Sulfanilic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)