Skip to Content
Merck
All Photos(1)

Documents

PHR1060

Supelco

Mesalamine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5-Aminosalicylic acid, 5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
2090421
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 1075
traceable to Ph. Eur. Y0000297
traceable to USP 1392705

API family

mesalamine, mesalazine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

275-280 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(O)c(c1)C(O)=O

InChI

1S/C7H7NO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,8H2,(H,10,11)

InChI key

KBOPZPXVLCULAV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Mesalamine is a drug, which is structurally similar to salicylates. It exhibits anti-inflammatory activity and is widely used as a medication for the treatment of inflammatory bowel disease, including ulcerative proctitis and Crohn′s disease.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Mesalamine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations using spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7173 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of UV spectrophotometric method for the determination of mesalamine in bulk and tablet formulation
Moharana KA, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 3(2), 19-21 (2011)
Spectrophotometric methods for the determination of mesalamine in bulk and pharmaceutical dosage forms
Acharjya KS, et al.
Journal of Pharmaceutical Education and Research, 1(1), 63-63 (2010)
Stephen J Goldie et al.
Cell death & disease, 9(11), 1072-1072 (2018-10-21)
Identifying soluble factors that influence epidermal integrity is critical for the development of preventative and therapeutic strategies for disorders such as ichthyosis, psoriasis, dermatitis and epidermal cancers. The transcription factor Grainyhead-like 3 (GRHL3) is essential for maintaining barrier integrity and
Alexander C Ford et al.
The American journal of gastroenterology, 106(12), 2070-2077 (2011-09-07)
Maintenance therapy with 5-aminosalicylates (5-ASAs) is recommended in patients with quiescent ulcerative colitis (UC), but compliance rates are low. Once-daily dosing may improve adherence, but impact on the relapse of disease activity is unclear as no previous meta-analysis has studied
A Lyakhovich et al.
Alimentary pharmacology & therapeutics, 31(2), 202-209 (2009-11-07)
Mesalazine (mesalamine) (5-ASA) is considered an anti-inflammatory drug for the treatment of inflammatory bowel disease. It is well tolerated by most patients and induces mucosal healing specifically in ulcerative colitis. Besides its anti-inflammatory properties, 5-ASA has been studied for cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service