Key Documents
RAD121
radiello Triangular Support Plate
pk of 20
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
packaging
pk of 20
manufacturer/tradename
Radiello Code 121
technique(s)
passive air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
A radiello diffusive sampler consists of 3 parts: the diffusive body, the adsorbent tube, and the triangular support plate. For personal air sampling, a vertical adapter can be installed. The adsorbent cartridge is removed from its glass or plastic storage tube and is inserted into the diffusive body. This is then threaded onto the triangular support plate in order to place the sampler in its sampling position.
The triangular support plate acts as both a closure and means of suspension for the Radiello diffusive body and cartridge adsorbent during passive air sampling. Each support plate is threaded for easy diffusive body attachment. Each plate includes a clip and transparent adhesive pocket to hold the Radiello barcode label.
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Study of the dispersion of VOCs emitted by a municipal solid waste landfill
Chiriac, R., et al.
Atmospheric Environment, 43 (11), 1926-1931 (2009)
Isabel M Morales Terrés et al.
Journal of environmental management, 91(12), 2754-2762 (2010-09-03)
This paper describes a novel methodology for evaluating the extent to which petrol stations affect their surroundings. The method is based on the fact that the ratio of the concentrations of aliphatic and aromatic hydrocarbon pollutants in the air of
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Anne Pennequin-Cardinal et al.
Talanta, 65(5), 1233-1240 (2008-10-31)
Radiello((R)) diffusive samplers filled with a thermally desorbable adsorbent (graphitised charcoal Carbograph 4) have been tested for the monitoring of BTEX. The sampling rates have been estimated under various controlled atmospheres in order to evaluate the effects of two factors
Rocco Rella et al.
The Science of the total environment, 487, 255-259 (2014-05-03)
The proofing treatment of the floor slabs with a solvent based material and successive treatment with epoxy resin resulted in significant air contamination in a primary school. The contamination caused illnesses in many occupants of school. The GC/MS investigations of
Articles
radiello® Diffusive Air Sampling Application - Ammonia
radiello® Diffusive Air Sampling Application - Ammonia
radiello® Diffusive Air Sampling Application - Hydrogen Sulfide
radiello® Diffusive Air Sampling Application - Hydrogen Sulfide
See All