RAD1202
radiello™ Diffusive Bodies
yellow, configured for reduced sampling rates, pk of 20
Synonym(s):
TM=["radiello"] Diffusive Bodies
About This Item
Recommended Products
material
polyethylene body
packaging
pk of 20
manufacturer/tradename
Radiello Code 120-2
technique(s)
passive air sampling: suitable
thickness × diffusive path length
5 mm × 150 mm
color
yellow
pore size
10 μm
application(s)
air monitoring
environmental
industrial hygiene
compatibility
configured for reduced sampling rates
Looking for similar products? Visit Product Comparison Guide
General description
Dimensions are 60 mm height x 16 mm diameter. There are four different diffusive bodies available. Each of which are application specific in design.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocols
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Related Content
Frequently Asked Questions - radiello® Products
Frequently Asked Questions - radiello® Products
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service