Skip to Content
Merck
All Photos(1)

Key Documents

RAD1203

radiello Diffusive Bodies

permeative, configured for anaesthetic gases and vapors, pk of 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

silicone body (strengthened by SS net)

packaging

pk of 20

manufacturer/tradename

Radiello Code 120-3

technique(s)

passive air sampling: suitable

thickness

50 μm

color

, permeative

application(s)

air monitoring
environmental
industrial hygiene

compatibility

configured for anaesthetic gases and vapors

Looking for similar products? Visit Product Comparison Guide

General description

The diffusive bodies are designed to house Radiello cartridge adsorbents during sampling. The diffusive bodies are threaded at one end for easy attachment to the Radiello triangular support plate. Unlike most of the cartridge adsorbents, the diffusive bodies are reusable and can be cleaned with a mild detergent between use. The diffusive bodies will collect dust (especially during outdoor sampling) and replacement is recommended after 4-5 uses.

Dimensions are 60 mm height x 16 mm diameter. There are four different diffusive bodies available. Each of which are application specific in design.

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
P Bruno et al.
Waste management (New York, N.Y.), 27(4), 539-544 (2006-05-23)
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic

Articles

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Protocols

Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.

Related Content

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service