Skip to Content
Merck
All Photos(1)

Key Documents

47168

Supelco

3,5-Di-tert-butyl-4-hydroxytoluene

analytical standard

Synonym(s):

3,5-Di-tert-butyl-4-hydroxytoluene, E321

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

room temp

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Gene Information

General description

3,5-Di-tert-4-butylhydroxytoluene belongs to the class of phenolic antioxidants, added to food and other products such as oils, fats and butter oil to prevent rancidity caused by the oxidation of unsaturated fats.

Application

3,5-Di-tert-4-butylhydroxytoluene may be used as an analytical reference standard for the quantification of the analyte in Eucalyptus wood using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra-high performance liquid chromatography coupled to mass spectrometry applied to the identification of valuable phenolic compounds from Eucalyptus wood
Santos.O.AS, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 938, 65-74 (2013)
Supercritical fluid extraction of phenolic compounds from Eucalyptus globulus Labill bark
Santos.O.AS, et al.
Journal of Supercritical Fluids, 71, 71-79 (2012)
Alessia Arena et al.
Food chemistry, 396, 133721-133721 (2022-07-24)
Phthalic acid esters (PAEs), known as phthalates, have a variety of industrial applications, mainly related to their ability to increase plastic softness,flexibility, and durability. With regard to toxicological aspects, several studies suggest that phthalates would act as endocrine-disrupting chemicals, and
E S Alinkina et al.
Prikladnaia biokhimiia i mikrobiologiia, 48(5), 564-569 (2012-10-30)
The antiradical properties of three samples of ginger (Zingiber officinale R.)-juice from fresh rhizome, essential oil, and extracts (oleoresin)-were studied and compared with the properties of synthetic antioxidant ionol (butylatedhydroxy-toluene, BHT). Reaction antioxidants with stable free 2,2-diphenyl-1-picrylhydrozyl radicals were used
Wageeh A Yehye et al.
Molecules (Basel, Switzerland), 17(7), 7645-7665 (2012-06-27)
A computer-aided predictions of antioxidant activities were performed with the Prediction Activity Spectra of Substances (PASS) program. Antioxidant activity of compounds 1, 3, 4 and 5 were studied using 1,1-diphenyl-2-picrylhydrazyl (DPPH) and lipid peroxidation assays to verify the predictions obtained

Articles

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service