Skip to Content
Merck
All Photos(2)

Documents

31519

Supelco

Ethoxyquin

PESTANAL®, analytical standard

Synonym(s):

1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO
CAS Number:
Molecular Weight:
217.31
Beilstein:
158223
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E324
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

1.03 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CCOc1ccc2NC(C)(C)C=C(C)c2c1

InChI

1S/C14H19NO/c1-5-16-11-6-7-13-12(8-11)10(2)9-14(3,4)15-13/h6-9,15H,5H2,1-4H3

InChI key

DECIPOUIJURFOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethoxyquin (EQ), an aromatic compound, is an antioxidant mostly used in animal and fish feeds. It is seen to inhibit carcinogenic effects of polycyclic aromatic hydrocarbons. Oxidation initiated by air, light or transition metals in unsaturated lipids can be stopped from propagation by EQ.

Application

EQ has been used as external standard in the determination of EQ and its major metabolite from Atlantic salmon tissues using HPLC with fluorescence detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Light sensitive: sensitive
Air sensitive: Handle under argon

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The inhibitory effects of ethoxyquin on the carcinogenic action of aflatoxin B1 in rats.
Cabral JR, Neal GE.
Cancer Letters, 19(2), 125-132 (1983)
Laia Sanchez Costa et al.
Analytical methods : advancing methods and applications, 12(32), 4080-4088 (2020-08-08)
Ethoxyquin (EQ) is a quinolone commonly used as an antioxidant additive and a fungicide. However, Regulation (EU) 2017/962 suspended its authorisation as a feed additive for all animal species and categories. The aim of this study is thus to ensure
Anup G Shah et al.
Water research, 39(17), 4251-4263 (2005-10-04)
The potential inhibitory effect of ethoxyquin, an antioxidant commonly used as a preservative in the food processing industry (e.g., for stabilizing dissolved air flotation residuals), was evaluated at concentrations up to 300 mg/L using a mixed, mesophilic (35 degrees C)
Alina Błaszczyk
Toxicology letters, 163(1), 77-83 (2005-11-04)
Ethoxyquin (1,2-dihydro-6-ethoxy-2,2,4-trimethylquinoline, EQ) is widely used in various food products and in animal feeds because of its powerful antioxidant activity. This compound was recently found to cause not only many unfavourable side-effects in animals fed with feeds containing it, but
J Andrew Keightley et al.
Molecular & cellular proteomics : MCP, 3(2), 167-175 (2003-12-17)
We are using a proteomic approach that combines two-dimensional electrophoresis and tandem mass spectrometry to detect and identify proteins that are differentially expressed in a cell line that is resistant to oxidative stress. The resistant cell line (OC14 cells) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service