Skip to Content
Merck
All Photos(1)

Key Documents

W218405

Sigma-Aldrich

Butylated hydroxytoluene

≥99%, FCC, FG

Synonym(s):

2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
FEMA Number:
2184
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
E Number:
E 321
NACRES:
NA.21
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1333/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.115

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99%

form

powder or crystals

autoignition temp.

878 °F

bp

265 °C (lit.)

mp

69-73 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

camphoraceous

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Butylated hydroxytoluene is a synthetic phenolic compound mainly used as an antioxidant and preservative in the food industry. It is used to prevent the lipid oxidation in oils and fat-containing foods.

Application


  • Role of odorant binding protein C12 in the response of Tribolium castaneum to chemical agents.: This article explores the impact of BHT on insect biochemistry, specifically how it affects the binding affinity of odorant-binding proteins in pests, providing insights into developing more effective pest control strategies (Wang et al., 2024).

  • A novel arylpiperazine derivative (LQFM181) protects against neurotoxicity induced by 3-nitropropionic acid in in vitro and in vivo models.: Research includes BHT as a comparative standard in studying neuroprotective agents, underscoring its role in biochemical pathways related to oxidative stress and neuroprotection (Campos et al., 2024).

  • Simultaneous Determination of Fipronil, Permethrin and their Key Related Substances in a Topical Drug Product by a Single Stability Indicating High Performance Liquid Chromatography Method.: BHT is used as an internal standard to assess the stability and integrity of pharmaceutical products, demonstrating its utility in pharmaceutical analysis (Rathnasekara et al., 2024).

  • Pyrano[2,3-c]pyrazole fused spirooxindole-linked 1,2,3-triazoles as antioxidant agents: Exploring their utility in the development of antidiabetic drugs via inhibition of a-amylase and DPP4 activity.: This study utilizes BHT as a reference antioxidant to compare the efficacy of newly synthesized compounds, providing a benchmark in antioxidant research for diabetes treatment (Chahal et al., 2024).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 327-330 (1997)
Silvia Carvajal et al.
Scientific reports, 9(1), 12848-12848 (2019-09-08)
Non-alcoholic fatty liver disease (NAFLD) is the most common cause of chronic liver disease worldwide, ranging from steatosis to non-alcoholic steatohepatitis (NASH). Recently, cerium oxide nanoparticles (CeO2NPs) have emerged as a new antioxidant agent with hepatoprotective properties in experimental liver
Safety assessment of butylated hydroxyanisole and butylated hydroxytoluene as antioxidant food additives
Williams GM, et al.
Food And Chemical Toxicology, 37(9), 1027-1038 (1999)
Safety aspects of food preservatives
Parke DV & Lewis DFV
Food Additives and Contaminants, 9(5), 561-577 (1992)
A M Vijesh et al.
European journal of medicinal chemistry, 46(11), 5591-5597 (2011-10-05)
In the present study two new series of Hantzsch 1,4-dihydropyridine derivatives (1,4-DHPs) containing substituted pyrazole moiety (4a-f and 5a-f) were synthesized by the reaction of 3-aryl-1H-pyrazole-4-carbaldehydes with 1,3-dicarbonylcompounds (ethylacetoacetate and methylacetoacetate) and ammonium acetate. The newly synthesized compounds were characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service