Skip to Content
Merck
All Photos(1)

Key Documents

Y2146

Sigma-Aldrich

Yeast Synthetic Drop-out Medium Supplements

without histidine, leucine, and tryptophan

Synonym(s):

Synthetic yeast drop-out mix, Yeast Synthetic Drop-out Media Supplements, Yeast media, amino acid dropout supplements

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

Quality Level

form

powder

technique(s)

transformation: suitable

application(s)

microbiology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component. Sigma′s Yeast Synthetic Drop-Out Media Supplements create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.
Yeast Synthetic Drop-out Medium Supplements are suitable for screening yeast transformants.

Other Notes

Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.

Preparation Note

Dissolve 1.46 g of supplement per liter of medium.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudeshna Nag et al.
Journal of cell science, 131(18) (2018-08-30)
Sorting endosomes (SEs) are the regulatory hubs for sorting cargo to multiple organelles, including lysosome-related organelles, such as melanosomes in melanocytes. In parallel, melanosome biogenesis is initiated from SEs with the processing and sequential transport of melanocyte-specific proteins toward maturing
Riddhi Atul Jani et al.
Journal of cell science, 128(17), 3263-3276 (2015-07-26)
Melanosomes are a class of lysosome-related organelles produced by melanocytes. Biogenesis of melanosomes requires the transport of melanin-synthesizing enzymes from tubular recycling endosomes to maturing melanosomes. The SNARE proteins involved in these transport or fusion steps have been poorly studied.

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service