Skip to Content
Merck
All Photos(1)

Key Documents

Y1375

Millipore

YPD Broth

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Yeast Peptone Dextrose Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

composition

Bacteriological peptone, 20 g/L
Glucose, 20 g/L
Yeast extract, 10 g/L

packaging

poly bottle of 1 kg
poly bottle of 250 g

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

color

beige

pH

6.3-6.7

application(s)

environmental
food and beverages

microbiology

suitability

nonselective for Candida spp.
nonselective for Pichia spp.
nonselective for Saccharomyces spp.
nonselective for Zygosaccharomyces spp.

General description

YPD Broth is a nutritious general liquid growth medium commonly used for the maintenance and propagation of Pichia pastoris, Saccharomyces cerevisiae, and other yeasts. It is a medium used for molecular cloning, protein expression, or as enrichment media. Peptic digest of animal tissue provides nitrogenous nutrients. Yeast extract provides nitrogenous nutrients as well as Vitamin B Complex. Dextrose provides the carbohydrate and energy source to support the growth of S. cerevisiae.

Application

YPD Broth is recommended for the growth of Saccharomyces cerevisiae for molecular biology purposes. This medium supports the growth of most heterotrophic microorganisms but due to their simple composition, they have been adopted as the basal media for the routine cultivation of yeasts. General methods in yeast genetics specify using Yeast Extract Peptone- Dextrose (YPD) medium for cultivating S. cerevisiae and other yeasts.

Quantity

50 g per liter

Preparation Note

1. Suspend 50 g in 1 L of distilled water.
2. Autoclave for 15 minutes at 121°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Christina Bergonzo et al.
Proteins, 88(9), 1189-1196 (2020-03-18)
ClpS2 is a small protein under development as a probe for selectively recognizing N-terminal amino acids of N-degron peptide fragments. To understand the structural basis of ClpS2 specificity for an N-terminal amino acid, all atom molecular dynamics (MD) simulations were
Runhui Liu et al.
Journal of the American Chemical Society, 136(11), 4333-4342 (2014-03-13)
Fungal infections are a major challenge to human health that is heightened by pathogen resistance to current therapeutic agents. Previously, we were inspired by host-defense peptides to develop nylon-3 polymers (poly-β-peptides) that are toxic toward the fungal pathogen Candida albicans
Veronique Archambault-Leger et al.
Biotechnology for biofuels, 5(1), 49-49 (2012-07-21)
The impact of hydrothermal flowthrough (FT) pretreatment severity on pretreatment and solubilization performance metrics was evaluated for three milled feedstocks (corn stover, bagasse, and poplar) and two conversion systems (simultaneous saccharification and fermentation using yeast and fungal cellulase, and fermentation
Ilias Kounatidis et al.
G3 (Bethesda, Md.), 8(5), 1637-1647 (2018-03-15)
Candida glabrata (C. glabrata) forms part of the normal human gut microbiota but can cause life-threatening invasive infections in immune-compromised individuals. C. glabrata displays high resistance to common azole antifungals, which necessitates new treatments. In this investigation, we identified five
Fatemeh Mohammadipanah et al.
Pharmaceutical biology, 56(1), 51-59 (2017-12-26)
Actinobacteria are a precious source of novel bioactive metabolites with potential pharmaceutical applications. Representatives of 11 genera of rare Actinobacteria were selected for the evaluation of antioxidant activity. Fermentation broths of the Actinobacteria were extracted and dosage of 10 to

Articles

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Protocols

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service