Skip to Content
Merck
All Photos(3)

Documents

G7021

Sigma-Aldrich

D-(+)-Glucose

powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Quality Level

Assay

≥99.5%

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

mp

150-152 °C (lit.)

solubility

H2O: 2g + 15 mL (Tested)

application(s)

agriculture

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

Glucose is the energy source of brain and of cells containing few or no mitochondria. It is also a substrate for anaerobic glycolysis. The human body stores glucose in the form of glycogen. Under low glucose conditions, it is released from liver and kidney via glycogen. Glucose is also required for the formation of di-, oligo- and polysaccharides.

Application

  • D-(+)-Glucose has been used for culturing of mammalian cells.
  • It has been used to study interactions between glucose and alanine and their influence on pentose phosphate pathway activity.
  • It has been used to perform glucose tolerance test in mice.

Other Notes

Mixed anomers

Preparation Note

The material may produce a yellowish solution if prepared at a higher concentration than 2g + 15ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferrier DR
Biochemistry (Lippincott's illustrated reviews) (2013)
Modulation of glucose-6-phosphate dehydrogenase, 6-phosphogluconate dehydrogenase and malic enzyme activity by glucose and alanine in Atlantic salmon, Salmo salar L. hepatocytes.
Sanden M et al.
Aquaculture (Amsterdam, Netherlands), 221, 469-480 (2003)
Ontogeny of circadian organization in the rat.
Yamazaki S et al.
Journal of biological rhythms, 24, 55-55 (2009)
A novel long term short interval physical activity regime improves body composition in mice.
Goh J and Ladiges WC
BMC Research Notes, 6, 66-66 (2013)
Jashanpreet Kaur et al.
PloS one, 10(3), e0122354-e0122354 (2015-03-27)
Genotyping studies of Australian Scedosporium isolates have revealed the strong prevalence of a recently described species: Scedosporium aurantiacum. In addition to occurring in the environment, this fungus is also known to colonise the respiratory tracts of cystic fibrosis (CF) patients.

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service