Skip to Content
Merck
All Photos(3)

Key Documents

XNAP2E

Sigma-Aldrich

Extract-N-Amp Plant PCR Kit

sufficient for 100 extractions, sufficient for 500 amplifications

Synonym(s):

Plant direct PCR kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55
Pricing and availability is not currently available.

usage

sufficient for 100 extractions
sufficient for 500 amplifications
sufficient for 500 reactions

Quality Level

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

colorless

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Extract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. A novel Extraction Solution eliminates the need for conventional freezing of plant tissues with liquid nitrogen, mechanical disruption, organic extraction, column purification, or precipitation of DNA. The kit also includes a PCR ReadyMix, especially formulated for amplification directly from extract.

Genomic DNA is extracted from 0.5 to 0.7 cm plant leaf disks that have been cut with a standard paper punch and simply incubated in Extraction Solution at 95 oC for 10 minutes. An equal volume of Dilution Solution is added to the extract to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and the Extract-N-Amp PCR ReadyMix, included in the kit.
The Extract-N-Amp Plant PCR Kits are intended for direct PCR (polymerase chain reaction). This kit contains all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. It is a novel Extraction Solution that eliminates the need for conventional freezing of plant tissues.
The PCR reaction mix facilitates amplification directly from the extract by using an antibody-based hot-start DNA polymerase for specific amplification.

Application

Extract-N-Amp Plant PCR Kit has been used to extract plant genomic DNA and in polymerase chain reaction (PCR).[1][2][3][4][5]

Extract-N-Amp Plant PCR Kit is suitable for use in genotyping.

It has been used forDNA extraction from fresh mushrooms.

Features and Benefits

  • Single-step extraction of plant genomic DNA for PCR in less than 15 minutes
  • A PCR ReadyMix, specially formulated for amplification directly from extract
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • Extract stable at 4 °C for at least 6 months
  • Plant genomic DNA can be extracted for PCR in under 15 minutes using a method.
  • No freezing, mechanical disruption, organic extraction, column purification or precipitation is necessary
  • A specially formulated PCR ReadyMix is provided for use with the extracted DNA
  • The Hot Start antibody ensures highly specific PCR amplification of genomic DNA
  • Suitable for high-throughput plant genetic analysis.
  • Lengthy enzymatic digestions are not required, saving time and effort.
  • The extracted DNA remains stable at 4 °C for atleast 6 months.

Components

Extract-N-Amp Plant PCR Kits contain Dilution solution, Extraction solution, and Extract-N-Amp PCR Reaction Mix (2X) (buffer, salts, dNTPs, Taq polymerase, and JumpStart Taq antibody)
Extraction Solution- E7526-12 mL

Dilution Solution- D5688-12mL

Extract-N-Amp PCR ReadyMix. This 2X PCR reaction mix contains buffer, salts, dNTPs, Taq polymerase, and JumpStart Taq antibody.- E3004- 5 x 1.2 ml

Collection tubes

Other Notes

The Extract-N-Amp Plant PCR Kits are for laboratory use only. Not for drug, household, or other uses.
The Extract-N-Amp Plant PCR Kits are for laboratory use only. Not for drug, household, or other uses.

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Len N Gillman et al.
Journal of evolutionary biology, 23(6), 1327-1330 (2010-05-12)
A faster rate of nuclear DNA evolution has recently been found for plants occupying warmer low latitudes relative to those in cooler high latitudes. That earlier study by our research group compared substitution rates within the variable internal transcribed spacer
S A Weller et al.
Journal of microbiological methods, 70(2), 379-383 (2007-06-26)
Real-time (TaqMan) PCR assays were developed to detect the strawberry angular leaf spot pathogen Xanthomonas fragariae (Xf) and the strawberry bacterial blight pathogen Xanthomonas arboricola pv. fragariae (Xaf). The Xf PCR (Xf gyrB) was designed within regions of the gyraseB
Sophie Lev et al.
Molecular plant-microbe interactions : MPMI, 22(9), 1093-1103 (2009-08-07)
Phosphorylated mitogen-activated protein kinases (MAPK) transmit signals by activation of their targets. The extent of signal transduction could depend on MAPK phosphorylation level, concentration, and subcellular localization. The pathogenicity MAPK Chk1 of the fungal corn pathogen Cochliobolus heterostrophus is required
Noa Eliahu et al.
Eukaryotic cell, 6(3), 421-429 (2007-01-24)
The maize pathogen Cochliobolus heterostrophus requires two mitogen-activated protein kinases (MAPKs), Chk1 and Mps1, to produce normal pigmentation. Young colonies of mps1 and chk1 deletion mutants have a white and autolytic appearance, which was partially rescued by a hyperosmotic environment.
Rena Shimizu et al.
Plant physiology, 149(2), 841-850 (2008-12-17)
The WUSCHEL-related homeobox (WOX) gene PRESSED FLOWER1 (PRS1) performs a conserved function during lateral organ development in Arabidopsis (Arabidopsis thaliana). Expressed in the periphery of the shoot meristem, PRS1 recruits founder cells that form lateral domains of vegetative and floral

Articles

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

See All

Protocols

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service