Skip to Content
Merck
All Photos(1)

Key Documents

T0826

Sigma-Aldrich

TBB

≥98% (HPLC), solid

Synonym(s):

4,5,6,7-Tetrabromo-2-azabenzimidazole, 4,5,6,7-Tetrabromobenzotriazole, NSC 231634, TBBt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6HN3Br4
CAS Number:
Molecular Weight:
434.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: 28 mg/mL

storage temp.

2-8°C

SMILES string

Brc1c(Br)c(Br)c2[nH]nnc2c1Br

InChI

1S/C6HBr4N3/c7-1-2(8)4(10)6-5(3(1)9)11-13-12-6/h(H,11,12,13)

InChI key

OMZYUVOATZSGJY-UHFFFAOYSA-N

Application

TBB was used to study casein kinase 2-dependent phosphorylation of DNA damage mediator protein MDC1.

Biochem/physiol Actions

TBB binds to the Val66 residue of casein kinase-2 and inhibits the binding of ATP/GTP. TBB is cell permeable; it induces caspase-dependent apoptosis and degrades hematopoietic lineage cell-specific protein 1 in Jurkat cells.
TBB is a highly selective, ATP/GTP-competitive inhibitor of casein kinase-2 (CK2) (IC50 = 900 nM and 1.6 mM, using rat liver and recombinant human CK2, respectively).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sarno et al.
FEBS letters, 496(1), 44-48 (2001-05-10)
The specificity of 4,5,6,7-tetrabromo-2-azabenzimidazole (TBB), an ATP/GTP competitive inhibitor of protein kinase casein kinase-2 (CK2), has been examined against a panel of 33 protein kinases, either Ser/Thr- or Tyr-specific. In the presence of 10 microM TBB (and 100 microM ATP)
J Ross Chapman et al.
EMBO reports, 9(8), 795-801 (2008-06-28)
Mammalian cells respond to DNA double-strand breaks (DSBs) by recruiting DNA repair and cell-cycle checkpoint proteins to such sites. Central to these DNA damage response (DDR) events is the DNA damage mediator protein MDC1. MDC1 interacts with several DDR proteins
Maria Ruzzene et al.
The Biochemical journal, 364(Pt 1), 41-47 (2002-05-04)
Incubation of Jurkat cells with 4,5,6,7-tetrabromobenzotriazole (TBB), a specific inhibitor of protein kinase CK2, induces dose-and time-dependent apoptosis as judged by several criteria. TBB-promoted apoptosis is preceded by inhibition of Ser/Thr phosphorylation of haematopoietic lineage cell-specific protein 1 (HS1) and
Andrew W Bergen et al.
PloS one, 10(7), e0126113-e0126113 (2015-07-02)
The Nicotine Metabolite Ratio (NMR, ratio of trans-3'-hydroxycotinine and cotinine), has previously been associated with CYP2A6 activity, response to smoking cessation treatments, and cigarette consumption. We searched for drug metabolizing enzyme and transporter (DMET) gene variation associated with the NMR
Kate J Treharne et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 24(5-6), 347-360 (2009-11-17)
Deletion of phenylalanine-508 (DeltaF508) from the first nucleotide-binding domain (NBD1) in the wild-type cystic fibrosis (CF) transmembrane-conductance regulator (wtCFTR) causes CF. However, the mechanistic relationship between DeltaF508-CFTR and the diversity of CF disease is unexplained. The surface location of F508

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service