Skip to Content
Merck
All Photos(4)

Documents

A2754

Sigma-Aldrich

Adenosine 5′-diphosphate sodium salt

bacterial, ≥95% (HPLC)

Synonym(s):

ADP sodium salt, ADP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O10P2
CAS Number:
Molecular Weight:
427.20
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

bacterial

Assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Na].Nc1ncnc2n(cnc12)C3OC(COP(O)(=O)OP(O)(O)=O)C(O)C3O

InChI

1S/C10H15N5O10P2/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(24-10)1-23-27(21,22)25-26(18,19)20/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H2,11,12,13)(H2,18,19,20)/t4-,6-,7-,10-/m1/s1

InChI key

XTWYTFMLZFPYCI-KQYNXXCUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Adenosine 5′-diphosphate (ADP) is an adenine nucleotide involved in energy storage and nucleic acid metabolism via its conversion into ATP by FOF1-ATP synthases. ADP can also be obtained by adenylate monophosphate (AMP) reaction catalyzed by adenylate kinase. ADP is a platelet agonist that is secreted from platelet dense granules. It promotes platelet activation through its interaction with ADP receptors P2Y1 and P2Y12. Upon conversion of ADP to adenosine by ecto-ADPases, platelet activation is inhibited via adenosine receptors.

Application

Adenosine 5′-diphosphate sodium salt has been used:
  • as a positive control of platelet aggregation to analyze antiplatelet activity of purified disintegrin by aggregometery.
  • in the preparation of stock solution for a mito stress test.
  • as a positive control in flow cytometry analysis to study the effect of pentameric C-reactive protein (pCRP) and monomeric CRP (mCRP) on the CD62p (P-selectin) expression of platelets.
  • as a platelet agonist to stimulate platelet aggregation.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryosuke Nakata et al.
Scientific reports, 7(1), 17170-17170 (2017-12-08)
Given the rising incidence of non-alcoholic fatty liver disease (NAFLD) in both adults and children, the development of a non-invasive diagnostic method for assessing disease progression to non-alcoholic steatohepatitis (NASH) has become an important research goal. Currently available non-invasive imaging
Kaushar Jahan et al.
Scientific reports, 10(1), 11603-11603 (2020-07-16)
Bone fracture repair is a multifaceted, coordinated physiological process that requires new bone formation and resorption, eventually returning the fractured bone to its original state. Currently, a variety of different approaches are pursued to accelerate the repair of defective bones
Glutathione peroxidase-3 deficiency promotes platelet-dependent thrombosis in vivo
Jin RC, et al.
Circulation, 123(18), 1963-1973 (2011)
Klaus H Nielsen et al.
RNA (New York, N.Y.), 15(1), 67-75 (2008-11-27)
The exon junction complex (EJC) is deposited onto spliced mRNAs and is involved in many aspects of mRNA function. We have recently reconstituted and solved the crystal structure of the EJC core made of MAGOH, Y14, the most conserved portion
Kaushar Jahan et al.
Carbohydrate polymers, 203, 60-70 (2018-10-16)
Biomineralization of soft scaffolds is a new venture in bone tissue engineering. This work aimed to develop a new injectable and in-situ gelling soft scaffold with chitosan and apatites through a rapid purine-crosslinking reaction. The scaffolds were fabricated by mixing

Protocols

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service