Skip to Content
Merck
All Photos(6)

Documents

L8000

Sigma-Aldrich

L-Leucine

≥98% (HPLC), suitable for ligand binding assays

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721722
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Leucine, reagent grade, ≥98% (HPLC)

grade

reagent grade

Assay

≥98% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

impurities

isoleucine and methionine, essentially free

color

white to off-white

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Leucine has been used as an agonist of mechanistic target of rapamycin (mTOR) to determine whether AMPK/mTOR pathway is involved in berberine-mediated autophagy in J774A.1 cells. It has also been used for the selection of transformed cells.

Biochem/physiol Actions

Leucine is a non-glucogenic, essential amino acid. It is a branched-chain amino acid that is a structural component of proteins. Leucine positively influences insulin release to eliminate toxic sugars out of the blood. The degradation of leucine leads to the formation of ketone bodies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dr. Victor Prisk
The Leucine Factor Diet: The Scientifically-Proven Approach to Combat Sugar, Burn Fat and Build Muscle (2015)
Berberine alleviates ox-LDL induced inflammatory factors by up-regulation of autophagy via AMPK/mTOR signaling pathway
Xiaodi Fan
Journal of Translational Medicine (2015)
Susan A Masino
Ketogenic Diet and Metabolic Therapies: Expanded Roles in Health and Disease (2016)
Markerless Gene Editing in the Hyperthermophilic Archaeon Thermococcus kodakarensis
Alexandra m
Bio-protocol (2017)
Sergio R Santa Maria et al.
Astrobiology (2020-01-07)
The biological risks of the deep space environment must be elucidated to enable a new era of human exploration and scientific discovery beyond low earth orbit (LEO). There is a paucity of deep space biological missions that will inform us

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service