Skip to Content
Merck
All Photos(2)

Key Documents

SML0053

Sigma-Aldrich

CP-101,606

≥98% (HPLC)

Synonym(s):

Traxoprodil; (1S,2S)-1-(4-hydroxy-phenyl)-2-(4-hydroxy-4-phenylpiperidino)-1-propanol; CP-101606

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO3
CAS Number:
Molecular Weight:
327.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +50 to +60° (c=1, MeOH)

color

white to tan

solubility

DMSO: ≥35 mg/mL

storage temp.

2-8°C

SMILES string

C[C@@H]([C@@H](O)c1ccc(O)cc1)N2CCC(O)(CC2)c3ccccc3

InChI

1S/C20H25NO3/c1-15(19(23)16-7-9-18(22)10-8-16)21-13-11-20(24,12-14-21)17-5-3-2-4-6-17/h2-10,15,19,22-24H,11-14H2,1H3/t15-,19+/m0/s1

InChI key

QEMSVZNTSXPFJA-HNAYVOBHSA-N

Gene Information

General description

CP-101,606 (Traxoprodil) is a substituted 4-phenylpiperidine and is localized in the fore brain neurons.

Application

CP-101,606 has been used as a N-methyl-D-aspartate (NMDA) receptor antagonist to study its role in recovery of spinal cord injuries.

Biochem/physiol Actions

CP-101,606 (Traxoprodil) plays a role in inhibiting glutamate-induced death in rats. It may exhibit therapeutic effects against human ischemia and neurodegenerative disorders. Traxoprodil is metabolized by cytochrome P450 (CYP) 2D6.
Traxoprodil (CP-101,606) is a potent noncompetitive antagonist of N-methyl-D-aspartate (NMDA) receptors, selective for the NR2B subunit. It has been shown to be neuroprotective in animal models of brain injury and stroke.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Becker et al.
Psychopharmacology, 236(12), 3451-3463 (2019-07-04)
The N-methyl-D-aspartate receptor (NMDAR) antagonist ketamine is known to have not only a rapid antidepressant effect but also dissociative side effects. Traxoprodil and lanicemine, also NMDA antagonists, are candidate antidepressant drugs with fewer side effects. In order to understand their
Chandra Prakash et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(8), 1350-1364 (2007-05-15)
Disposition of traxoprodil ({1-[2-hydroxy-2-(4-hydroxy-phenyl)-1-methyl-ethyl]-4-phenyl-piperidin-4-ol}mesylate; TRX), a selective antagonist of the N-methyl-d-aspartate class of glutamate receptors, was investigated in rats and dogs after administration of a single i.v. bolus dose of [(14)C]TRX. Total mean recoveries of the radiocarbon were 92.5 and
Timothy J Taylor et al.
Clinical pharmacokinetics, 45(10), 989-1001 (2006-09-21)
Traxoprodil, a substituted 4-phenylpiperidine, is an N-methyl-D-aspartate (NMDA) receptor antagonist that is selective for receptors containing the NR2B subunit. In vivo and in vitro studies examining the disposition of traxoprodil have demonstrated that it is mainly metabolised by cytochrome P450
Weronika Stasiuk et al.
Journal of neural transmission (Vienna, Austria : 1996), 124(3), 387-396 (2016-12-03)
Pre-clinical and clinical studies indicated that a blockade of the NMDA receptor complex creates new opportunities for the treatment of affective disorders, including depression. The aim of the present study was to assess the influence of traxoprodil (10 mg/kg) on the
Bo Chen et al.
Cell, 174(3), 521-535 (2018-07-24)
Many human spinal cord injuries are anatomically incomplete but exhibit complete paralysis. It is unknown why spared axons fail to mediate functional recovery in these cases. To investigate this, we undertook a small-molecule screen in mice with staggered bilateral hemisections

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service