Skip to Content

D054

R(+)-SCH-23390 hydrochloride

≥98% (HPLC), dopamine receptor antagonist, solid

Synonym(s):

R(+)-7-Chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H18ClNO · HCl
CAS Number:
Molecular Weight:
324.24
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

R(+)-SCH-23390 hydrochloride, ≥98% (HPLC), solid

Quality Segment

assay

≥98% (HPLC)

form

solid

optical activity

[α]/D +32.1°, c = 1 in DMF(lit.)

storage condition

desiccated, protect from light

color

white

solubility

H2O: >5 mg/mL

SMILES string

Cl[H].CN1CCc2cc(Cl)c(O)cc2[C@H](C1)c3ccccc3

InChI

1S/C17H18ClNO.ClH/c1-19-8-7-13-9-16(18)17(20)10-14(13)15(11-19)12-5-3-2-4-6-12;/h2-6,9-10,15,20H,7-8,11H2,1H3;1H/t15-;/m1./s1

InChI key

OYCAEWMSOPMASE-XFULWGLBSA-N

Gene Information

human ... DRD1(1812)

Application

R(+)-SCH-23390 hydrochloride has been used:
  • for intraperitoneal injection into experimental rats to study the role of D1 receptor in sensitization
  • as a D1 receptor-selective antagonist for intracranial drug infusions
  • as an antagonist of D1 receptor to study its effect on learning ability in fish

Biochem/physiol Actions

R(+)-SCH-23390 hydrochloride is a selective D1 dopamine receptor antagonist. It has greater binding capacity towards 5-hydroxytryptamine 2 (5-HT2) and 5-HT1C serotonin receptor subtypes in in vitro. R(+)-SCH-23390 has a half-life of about 25 minutes after administration. It is known to possess anti-stereotypic effect and cataleptogenic effect. It also has inhibitory effect on motor activity.

Features and Benefits

This compound is featured on the Dopamine Receptors and Potassium Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Sold with the permission of Schering-Plough.


Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library