Skip to Content
Merck
All Photos(1)

Key Documents

SML1899

Sigma-Aldrich

SR144528

≥98% (HPLC)

Synonym(s):

5-(4-Chloro-3-methylphenyl)-1-[(4-methylphenyl)methyl]-N-[(1S,2S,4R)-1,3,3-trimethylbicyclo[2.2.1]hept-2-yl]-1H-pyrazole-3-carboxamide, SR-144528

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H34ClN3O
CAS Number:
Molecular Weight:
476.05
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@]12[C@H](NC(C3=NN(CC4=CC=C(C)C=C4)C(C5=CC=C(Cl)C(C)=C5)=C3)=O)C(C)(C)[C@@H](C2)CC1

InChI

1S/C29H34ClN3O/c1-18-6-8-20(9-7-18)17-33-25(21-10-11-23(30)19(2)14-21)15-24(32-33)26(34)31-27-28(3,4)22-12-13-29(27,5)16-22/h6-11,14-15,22,27H,12-13,16-17H2,1-5H3,(H,31,34)/t22-,27-,29+/m1/s1

InChI key

SUGVYNSRNKFXQM-XRHWURSXSA-N

Application

SR144528 has been used as a cannabinoid CB2 receptor antagonist:
  • to study its blocking effect on the anti-dyskinetic functionality of HU-308
  • to study its effects on spontaneous excitatory postsynaptic currents (sEPSCs) from the globus pallidus neurons
  • to analyze its effect on the gastric emptying in rats

Biochem/physiol Actions

SR144528 is a potent and highly selective cannabinoid CB2 receptor inverse agonist.
SR144528 is a potent and highly selective cannabinoid CB2 receptor inverse agonist. SR144528 exhibited a Ki of 0.6 nM at CB2 compared to 400 nM at the related CB1 receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Tan et al.
Brain research, 1654(Pt A), 24-33 (2016-10-23)
Fibrosis in ventricular system has a role in hydrocephalus following intraventricular hemorrhage (IVH). The cannabinoid receptor 2 (CB2) has been reported to participate in alleviating the fibrosis process of many diseases. However, its role in fibrosis after IVH was unclear
M Rinaldi-Carmona et al.
The Journal of pharmacology and experimental therapeutics, 284(2), 644-650 (1998-03-07)
Based on both binding and functional data, this study introduces SR 144528 as the first, highly potent, selective and orally active antagonist for the CB2 receptor. This compound which displays subnanomolar affinity (Ki = 0.6 nM) for both the rat
A A Izzo et al.
Naunyn-Schmiedeberg's archives of pharmacology, 360(2), 221-223 (1999-09-24)
We have studied the effect of WIN 55,212-2 (a psychoactive cannabinoid agonist), cannabinol (a nonpsychoactive cannabinoid agonist), SR141716A, a cannabinoid CB1 antagonist, and SR144528, a cannabinoid CB2 antagonist, on gastric emptying in the rat. WIN 55,212-2 (0.1-5 mg/kg, i.p.) and
I Freiman et al.
Neuroscience, 133(1), 305-313 (2005-05-17)
The globus pallidus receives its major glutamatergic input from the subthalamic nucleus and subthalamic nucleus neurons synthesize CB1 cannabinoid receptors. The hypothesis of the present work was that CB1 receptors are localized in terminals of subthalamo-pallidal glutamatergic axons and that
Peggy Rentsch et al.
Neurobiology of disease, 134, 104646-104646 (2019-11-02)
L-dopa induced dyskinesia (LID) is a debilitating side-effect of the primary treatment used in Parkinson's disease (PD), l-dopa. Here we investigate the effect of HU-308, a cannabinoid CB2 receptor agonist, on LIDs. Utilizing a mouse model of PD and LIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service