Skip to Content
Merck
All Photos(1)

Key Documents

SCP0060

Sigma-Aldrich

CGRP II 8-37 human / β CGRP 8-37 human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C138H229N43O38S1
Molecular Weight:
3130.63
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Met-Val-Lys-Ser-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2

Application

β-Calcitonin gene related peptide (β-CGRP) regulates cardiovascular homeostasis and nociception via G-coupled protein receptor (GCPR) calcitonin receptor-like receptor (CALCRL).
CGRP II 8-37 is used as a CGRP receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joseph J Lynch et al.
European journal of pharmacology, 623(1-3), 96-102 (2009-09-22)
The triptans, serotonin 5-HT(1B/1D) receptor agonists exemplified by sumatriptan, are a mainstay migraine therapy but have class labeling contraindicating their use in patients with coronary artery disease. Triptans constrict human coronary artery in vitro, and there are case reports of
Vince T Nguyen et al.
Peptides, 33(1), 77-82 (2011-11-24)
Cardiac epinephrine and calcitonin gene-related peptide (CGRP) are produced by intrinsic cardiac adrenergic cells (ICA cells) residing in human and animal hearts. ICA cells are neuroparicine cells expressing δ-opioid receptors (DOR). We hypothesized that δ-opioid stimulation of ICA cells enhances
Toomas Talme et al.
Journal of neuroimmunology, 196(1-2), 60-66 (2008-04-22)
CGRP significantly stimulated migration of non-activated and anti-CD3 activated T lymphocytes into a collagen matrix when present inside the collagen, whereas somatostatin-14, NPY, substance P, VIP, beta-endorphin and metenkephalin had no or little effect. The CGRP antagonist CGRP 8-37 abrogated
Paris Avgoustou et al.
ACS pharmacology & translational science, 3(4), 706-719 (2020-08-25)
The hormone adrenomedullin has both physiological and pathological roles in biology. As a potent vasodilator, adrenomedullin is critically important in the regulation of blood pressure, but it also has several roles in disease, of which its actions in cancer are
I-Ching Hou et al.
Peptides, 30(12), 2228-2232 (2009-09-02)
We found that beta-lactotensin (His-Ile-Arg-Leu), which has been isolated as an ileum-contracting peptide from chymotrypsin digest of bovine beta-lactoglobulin, dose-dependently suppresses food intake after intracerebroventricular (i.c.v.) or intraperitoneal administration at a dose of 40 nmol/mouse or 100mg/kg, respectively, in fasted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service