Skip to Content
Merck

C8198

Anti-Calcitonin Gene Related Peptide antibody produced in rabbit

whole antiserum

Synonym(s):

CGRP Antibody - Anti-Calcitonin Gene Related Peptide antibody produced in rabbit, Cgrp Antibody, Anti-CGRP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
Citations:
189
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

rat

packaging

antibody small pack of 25 μL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Calca(24241)

General description

Calcitonin Gene Related Peptide (CGRP), a cyclic peptide, belongs to a small family of peptides encoded by the calcitonin/CGRP gene complex. The calcitonin/CGRP gene complex consists of at least two genes, encoding for two forms of CGRP: -CGRP (CGRP-I) and -CGRP (CGRP-II). CGRP is synthesized and co-localized with calcitonin in the parafollicular C-cells of the thyroid gland and medullary thyroid carcinoma. It is principally considered a neuropeptide widely distributed in the central and peripheral nervous systems.

Immunogen

CGRP-KLH

Application

Anti-Calcitonin Gene Related Peptide antibody produced in rabbit has been used in:
  • dot blot immunoassay
  • fluorescent labeling
  • confocal microscopy
  • immunohistofluorescence
  • epifluorescence imaging

Anti-Calcitonin Gene Related Peptide antibody produced in rabbit was used for immunohistochemistry of trigeminal ganglia cell cultures at a dilution of 1:200 and for immunocytochemistry of mouse lung slices at a dilution of 1:500.

Biochem/physiol Actions

Anti-Calcitonin Gene Related Peptide reacts with CGRP (rat). Cross-reactivity is observed with CGRP (human) and β-CGRP (human).
CGRP is thought to act as a neurotransmitter or neuromodulator, particularly in the regulation of autonomic (e.g., cardiovascular, respiratory, gastrointestinal, taste, food intake and sleep) and limbic functions. CGRP is probably involved with substance P in mediating neurogenic inflammation and transmission of pain stimuli via the peripheral nervous system.
Calcitonin Gene Related Peptide is a G protein-coupled receptor that binds receptor-activity-modifying proteins (RAMP)1, -2 and -3. CLR has been implicated in conditions such as obesity, diabetes, osteoporosis, migraine and cardiovascular disease.

Physical form

Supplied as a liquid containing 0.1% sodium azide as a preservative

Preparation Note

For continuous use, store 2-8°C. For extended storage, solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free"freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.
delipidized

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Yae K Sugimura et al.
Journal of neurophysiology, 115(6), 2721-2739 (2016-02-19)
A large majority of neurons in the superficial layer of the dorsal horn projects to the lateral parabrachial nucleus (LPB). LPB neurons then project to the capsular part of the central amygdala (CeA; CeC), a key structure underlying the nociception-emotion
Lina Almarestani et al.
Molecular pain, 4, 56-56 (2008-11-19)
Pain is a major symptom associated with chronic inflammation. In previous work from our laboratory, we have shown that in animal models of neuropathic pain there is a sprouting of sympathetic fibers into the upper dermis, a territory normally devoid
Kumiko Ishida et al.
Anesthesiology, 121(5), 1068-1079 (2014-07-06)
The aim of this study was to clarify the roles of calcitonin gene-related peptide (CGRP) in postoperative pain and inflammatory pain. αCGRP knockout mice that the authors have developed and wild-type mice were used. Pain behaviors were assessed after incision



Global Trade Item Number

SKUGTIN
C8198-.2ML04061833521908
C8198-25UL04061838357656