Merck
  • Discovery of a First-in-Class Potent Small Molecule Antagonist against the Adrenomedullin-2 Receptor.
Discovery of a First-in-Class Potent Small Molecule Antagonist against the Adrenomedullin-2 Receptor.
ACS pharmacology & translational science (2020-08-25)
Paris Avgoustou, Ameera B A Jailani, Jean-Olivier Zirimwabagabo, Matthew J Tozer, Karl R Gibson, Paul A Glossop, James E J Mills, Roderick A Porter, Paul Blaney, Peter J Bungay, Ning Wang, Alice P Shaw, Kamilla J A Bigos, Joseph L Holmes, Jessica I Warrington, Timothy M Skerry, Joseph P A Harrity, Gareth O Richards
ABSTRACT
The hormone adrenomedullin has both physiological and pathological roles in biology. As a potent vasodilator, adrenomedullin is critically important in the regulation of blood pressure, but it also has several roles in disease, of which its actions in cancer are becoming recognized to have clinical importance. Reduced circulating adrenomedullin causes increased blood pressure but also reduces tumor progression, so drugs blocking all effects of adrenomedullin would be unacceptable clinically. However, there are two distinct receptors for adrenomedullin, each comprising the same G protein-coupled receptor (GPCR), the calcitonin receptor-like receptor (CLR), together with a different accessory protein known as a receptor activity-modifying protein (RAMP). The CLR with RAMP2 forms an adrenomedullin-1 receptor, and the CLR with RAMP3 forms an adrenomedullin-2 receptor. Recent research suggests that a selective blockade of adrenomedullin-2 receptors would be therapeutically valuable. Here we describe the design, synthesis, and characterization of potent small-molecule adrenomedullin-2 receptor antagonists with 1000-fold selectivity over the adrenomedullin-1 receptor, although retaining activity against the CGRP receptor. These molecules have clear effects on markers of pancreatic cancer progression in vitro, drug-like pharmacokinetic properties, and inhibit xenograft tumor growth and extend life in a mouse model of pancreatic cancer. Taken together, our data support the promise of a new class of anticancer therapeutics as well as improved understanding of the pharmacology of the adrenomedullin receptors and other GPCR/RAMP heteromers.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
CGRP II 8-37 human / β CGRP 8-37 human
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture