Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502012

Sigma-Aldrich

Anti-INOS antibody produced in rabbit

affinity isolated antibody

Synonym(s):

INDUCIBLE, MACROPHAGE, NITRIC OXIDE SYNTHASE, NOS2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 131 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOS2(4843)

General description

Anti-INOS antibody detects endogenous levels of total INOS protein.
The INOS (inducible nitric oxide synthase) gene is mapped to human chromosome 17q11.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human iNOS.

Immunogen Range: 1101-1150

Application

Anti-INOS, C-Terminal antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Upregulation of INOS (inducible nitric oxide synthase) is observed during infections, tissue injury, and inflammation. INOS is responsible to produce NO (nitric oxide), which controls blood flow by inducing vascular relaxation. NO acts as a neurotransmitter and can facilitate immune responses. NO can also bring about anti-thrombotic and anti-inflammatory effects in response to oxidative stress, hypoxia, inflammatory cytokines (TNF-α, IL-1β, IFN-γ), and lipopolysaccharide endotoxin. iNOS generates more NO than the neuronal and endothelial NOS. Unlike the two isoforms, iNOS is calcium independent. INOS might be associated with type 2 diabetes mellitus.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nora Kariche et al.
Journal of oncology, 2019, 5649846-5649846 (2019-12-31)
Laryngeal squamous cell carcinoma (LSCC) is an aggressive malignancy which lacks early predictors of prognosis. Here, we hypothesized that expression and prognostic characterization of the critical mediators of epithelial to mesenchymal transition (EMT) may provide key information in this regard.
Shu-Jyuan Chang et al.
Diagnostics (Basel, Switzerland), 10(5) (2020-05-01)
Glioma is characterized by a high heterogeneity in the brain tumor. Abundant tumor-associated macrophages (TAMs) exist as neoplastic tissues, implicating tumor plasticity and thus leading to therapeutic challenges. Vascular adhesion protein (VAP-1) potentially serves as a mediator for TAM immunity
Jin Li et al.
Molecular medicine reports, 16(4), 4253-4258 (2017-07-22)
Isorhamnetin (ISH) is a flavonoid primarily obtained from the fruit of Hippophae rhamnoides L., which possesses anti‑inflammatory properties. However, the effect of ISH on the expression of inflammatory mediators in response to interleukin (IL)‑1β stimulation has not been elucidated. The
Sudeshna Das et al.
Journal of neuroimmunology, 313, 161-175 (2017-11-18)
Over activation of glial cell derived innate immune factors induces neuro-inflammation that results in neurodegenerative disease, like working memory impairment. In this study, we have investigated the role of andrographolide, a major constituent of Andrographis paniculata plant, in reduction of
Rajendran Jayaganesh et al.
Journal of biochemical and molecular toxicology, 34(3), e22441-e22441 (2020-01-12)
Inflammation plays a vital role in the process of carcinogenesis and anti-inflammatory properties of phytochemicals are gaining more attention in the chemoprevention of cancer. The present study was designed to evaluate the anti-inflammatory potential of citronellol (CT) on 7,12-dimethylbenz(a)anthracene (DMBA)-induced mammary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service